Skip to Content
Merck
All Photos(2)

Key Documents

GF13770637

Zinc

foil, 50x50mm, thickness 0.1mm, as rolled, 99.95+%

Synonym(s):

Zinc, ZN000250

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Zn
CAS Number:
Molecular Weight:
65.39
MDL number:
UNSPSC Code:
12141750
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

1 mmHg ( 487 °C)

Assay

99.95%

form

foil

manufacturer/tradename

Goodfellow 137-706-37

resistivity

5.8 μΩ-cm, 20°C

size × thickness

50 x 50 mm × 0.1 mm

bp

907 °C (lit.)

mp

420 °C (lit.)

density

7.133 g/mL at 25 °C (lit.)

SMILES string

[Zn]

InChI

1S/Zn

InChI key

HCHKCACWOHOZIP-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A new vapor-phase hydrothermal method to concurrently grow ZnO nanotube and nanorod array films on different sides of a zinc foil substrate.
Haimin Zhang et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 18(17), 5165-5169 (2012-03-21)
Conghua Lu et al.
Chemical communications (Cambridge, England), (33)(33), 3551-3553 (2006-08-22)
Large-scale, ultralong ZnO nanowire and nanobelt arrays with honeycomb-like micropatterns have been fabricated by hydrothermal oxidation of zinc foil in aqueous alkaline (NH4)2S2O8 solutions.
Rui Gao et al.
Nanoscale, 5(5), 1894-1901 (2013-01-29)
In this paper, ZnO nanorod-nanosheet hierarchical structures were fabricated using a facile method on zinc foil and used as flexible photoanodes in dye-sensitized solar cells (DSCs). Compared to nanorods (NRs) obtained by the dissolution-precipitation method, the nanorod-nanosheet (NR-NS) hierarchical structures
Tandra Ghoshal et al.
Nanotechnology, 19(6), 065606-065606 (2008-02-13)
ZnO nanowire arrays were synthesized on zinc foil by a simple thermal evaporation process at relatively low temperature. Morphology and size controlled synthesis of the ZnO nanostructures was achieved by variation of the synthesis temperature, reaction time and the surface
Zhonghao Li et al.
Chemical communications (Cambridge, England), (41)(41), 6273-6275 (2009-10-15)
Aligned ZnO arrays with branched rods, hexagonal rods, quasi-round rods, hexagonal pyramids and porous nanostructures are constructed on zinc foil in the ionic liquid tetrabutylammonium hydroxide aqueous solution.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service