Skip to Content
Merck
All Photos(2)

Key Documents

GF09534359

Indium

foil, 150x150mm, thickness 0.20mm, as rolled, 99.999%

Synonym(s):

Indium, IN000260

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
In
CAS Number:
Molecular Weight:
114.82
MDL number:
UNSPSC Code:
12141719
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

<0.01 mmHg ( 25 °C)

Assay

≥99.999%

form

foil

manufacturer/tradename

Goodfellow 095-343-59

resistivity

8.37 μΩ-cm

mp

156.6 °C (lit.)

density

7.3 g/mL at 25 °C (lit.)

SMILES string

[In]

InChI

1S/In

InChI key

APFVFJFRJDLVQX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anatomic repair of internal perforations with indium foil and silver amalgam: outline of a method.
W P Auslander et al.
The New York journal of dentistry, 39(10), 454-457 (1969-12-01)
M L Arnold et al.
Medical physics, 29(11), 2718-2724 (2002-12-05)
Manganese (Mn) is an essential nutrient for growth and development. Unfortunately, overexposure can lead to neurological damage, which is manifested as a movement disorder marked by tremors. Preclinical symptoms have been found in populations occupationally exposed to the element, and
Mizuho Fushitani et al.
Optics express, 19(10), 9600-9606 (2011-06-07)
Generation of single-order laser harmonics in extreme ultraviolet (EUV) and the application to the time-resolved photoelectron spectroscopy of I(2) are demonstrated. The EUV pulses at 80 nm were generated from Kr as the 5th order harmonics of intense 400 nm
C Takada et al.
Radiation protection dosimetry, 144(1-4), 575-579 (2010-11-11)
At the Nuclear Fuel Cycle Engineering Laboratories of the Japan Atomic Energy Agency (JAEA), small pieces of indium foil incorporated into personal dosemeters have been used for personnel screening in criticality accidents. Irradiation tests of the badges were performed using
Loïc Bertrand et al.
The Analyst, 138(16), 4463-4469 (2013-04-05)
In order to fully characterize the zinc white artists' pigment (ZnO), much used since the mid-nineteenth century, three samples collected in the early 20th century were studied using a combination of synchrotron and macroscopic photoluminescence spectroscopy and imaging. An improved

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service