Skip to Content
Merck
All Photos(2)

Key Documents

GF00832081

Manganese

foil, not light tested, 50x50mm, thickness 0.015mm, permanent polyester support, 98.7%

Synonym(s):

Manganese, MN000170

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Mn
CAS Number:
Molecular Weight:
54.94
MDL number:
UNSPSC Code:
12141724
PubChem Substance ID:
NACRES:
NA.23

Assay

≥98.7%

form

foil

manufacturer/tradename

Goodfellow 008-320-81

resistivity

185 μΩ-cm, 20°C

L × W × thickness

50 mm × 50 mm × 0.015 mm

bp

1962 °C (lit.)

mp

1244 °C (lit.)

density

7.3 g/mL at 25 °C (lit.)

SMILES string

[Mn]

InChI

1S/Mn

InChI key

PWHULOQIROXLJO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Monika Meyer-Baron et al.
Neurotoxicology, 36, 1-9 (2013-02-20)
Results from a meta-analysis of aggregated data provoked a new analysis using individual data on the neuropsychological performance of occupationally exposed workers. Data from eight studies examining 579 exposed and 433 reference participants were included, 28 performance variables analyzed. The
Pattama Saisaha et al.
Chemical Society reviews, 42(5), 2059-2074 (2012-12-15)
The development of new catalytic systems for cis-dihydroxylation and epoxidation of alkenes, based on atom economic and environmentally friendly concepts, is a major contemporary challenge. In recent years, several systems based on manganese catalysts using H(2)O(2) as the terminal oxidant
Håkan Nilsson et al.
Biochimica et biophysica acta, 1837(8), 1257-1262 (2014-04-15)
The binding affinity of the two substrate-water molecules to the water-oxidizing Mn₄CaO₅ catalyst in photosystem II core complexes of the extremophilic red alga Cyanidioschyzon merolae was studied in the S₂ and S₃ states by the exchange of bound ¹⁶O-substrate against
R Allemang-Grand et al.
Neurobiology of aging, 36(2), 638-647 (2014-10-15)
Murine models of Alzheimer's disease (AD) have been used to draw associations between atrophy of neural tissue and underlying pathology. In this study, the early-onset TgCRND8 mouse model of AD and littermate controls were scanned longitudinally with in vivo manganese-enhanced
Mirna Lechpammer et al.
Annals of neurology, 75(4), 608-612 (2014-03-07)
We followed a patient with manganese transporter deficiency due to homozygous SLC30A10 mutations from age 14 years until his death at age 38 years and present the first postmortem findings of this disorder. The basal ganglia showed neuronal loss, rhodanine-positive

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service