G11004
Guaiazulene
99%
Synonym(s):
1,4-Dimethyl-7-isopropylazulene
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C15H18
CAS Number:
Molecular Weight:
198.30
Beilstein:
1365001
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
99%
bp
153 °C/7 mmHg (lit.)
mp
27-29 °C (lit.)
density
0.976 g/mL at 25 °C (lit.)
SMILES string
CC(C)c1ccc(C)c2ccc(C)c2c1
InChI
1S/C15H18/c1-10(2)13-7-5-11(3)14-8-6-12(4)15(14)9-13/h5-10H,1-4H3
InChI key
FWKQNCXZGNBPFD-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Guaiazulene can be used as a starting material for the synthesis of:
- Azulene-based dye molecules such as 3-(7-isopropyl-1,4-dimethylazulen-3-yl)-2-cyanoacrylic acid and 5-(7-isopropyl-1,4-dimethylazulen-3-yl)-2-cyanopenta-2,4-dienoic acid.
- Stilbazulenyl nitrone, a second-generation azulenyl nitrone which can be used as a chain-breaking antioxidant.
- Bis-azulenyl based near-infrared fluorescence quencher.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
235.4 °F - closed cup
Flash Point(C)
113 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Felix Momm et al.
Strahlentherapie und Onkologie : Organ der Deutschen Rontgengesellschaft ... [et al], 179(10), 708-712 (2003-10-21)
Radiation treatment may induce acute skin reactions. There are several methods of managing them. Validity of these methods, however, is not sufficiently studied. We therefore investigated, whether moist skin care with 3% urea lotion will reduce acute radiation skin toxicity.
Dawei Chen et al.
Journal of agricultural and food chemistry, 60(1), 112-123 (2011-12-14)
Fifteen new guaiazulene-based terpenoids designated anthogorgienes A-O (1-15) were isolated from a Chinese gorgonian Anthogorgia sp., together with eight known analogues (16-23). The structural patterns were classified into monomers, dimers, and trimers, which were supposed to be generated from a
Allergic contact cheilitis to guaiazulene.
G Angelini et al.
Contact dermatitis, 10(5), 311-311 (1984-05-01)
An Azulene Dimer as a Near-Infrared Quencher.
Pham W, et al.
Angewandte Chemie (International Edition in English), 41(19), 3659-3662 (2002)
Lei Wang et al.
Mutation research, 530(1-2), 19-26 (2003-10-18)
The photomutagenicity of the popular skin conditioning agents azulene and guaiazulene were tested in Salmonella typhimurium TA98, TA100 and TA102. Following irradiation with UVA and/or visible light, both azulene and guaiazulene exhibited mutagenicity 4-5-fold higher than the spontaneous background mutation.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service