All Photos(3)
About This Item
Linear Formula:
C2H5NHCONH2
CAS Number:
Molecular Weight:
88.11
Beilstein:
1740512
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
97%
mp
87-90 °C (lit.)
SMILES string
CCNC(N)=O
InChI
1S/C3H8N2O/c1-2-5-3(4)6/h2H2,1H3,(H3,4,5,6)
InChI key
RYECOJGRJDOGPP-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
200.1 °F - closed cup
Flash Point(C)
93.4 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Shinichi Kawata et al.
Anatomical science international, 94(2), 209-215 (2019-01-04)
Formaldehyde (FA) is frequently used to embalm human cadavers that are employed to teach gross anatomy to medical and dental students. However, exposure to FA is harmful to both students and educators. The aim of this study was to reduce
I E Dreosti et al.
Research communications in chemical pathology and pharmacology, 41(2), 265-281 (1983-08-01)
N-Nitrosoethylurea (ENU) and ethylurea (EU) with sodium nitrite (NaNO2) were administered daily by mouth to Hooded Wistar rats. Doses ranged from 1-50 mg ENU/kg body weight (day-1) and from 20-70 mg EU/kg body weight day-1 and extended over days 7
H Suzuki et al.
Chemical & pharmaceutical bulletin, 45(10), 1688-1693 (1997-11-14)
The most prevalent means for producing solid dispersions of nifedipine, a poorly water-soluble drug, are the solvent based processes that bring problems of environmental and health. We have investigated the preparation of solid dispersions of nifedipine (mp 173 degrees C)
The C-terminal extrahelical peptide of type I collagen and its role in fibrillogenesis in vitro: effects of ethylurea.
M J Capaldi et al.
Biopolymers, 23(2), 313-323 (1984-02-01)
H Inoue et al.
Mutation research, 88(3), 281-290 (1981-03-01)
The standard sex-linked recessive lethal test was used to test whether NO2 induces lethal mutations in male germ cells of Drosophila in the presence or absence of alkylureas. Methylurea, ethylurea and NO2 alone did not enhance the mutation frequency significantly.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service