Skip to Content
Merck
All Photos(3)

Key Documents

E40609

Sigma-Aldrich

Ethyl nicotinate

99%

Synonym(s):

Nicotinic acid ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H9NO2
CAS Number:
Molecular Weight:
151.16
Beilstein:
122937
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

liquid

refractive index

n20/D 1.504 (lit.)

bp

223-224 °C (lit.)

mp

8-10 °C (lit.)

density

1.107 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)c1cccnc1

InChI

1S/C8H9NO2/c1-2-11-8(10)7-4-3-5-9-6-7/h3-6H,2H2,1H3

InChI key

XBLVHTDFJBKJLG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

199.4 °F - closed cup

Flash Point(C)

93 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ibrahim Uçar et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 72(1), 11-16 (2008-11-18)
Mononuclear copper(II) saccharinate (sac) complex containing ethylnicotinate (enc), [Cu(enc)(2)(sac)(2)(H(2)O)].1.4H(2)O has been synthesized and characterized by spectroscopic (IR, UV-vis, EPR), X-ray diffraction technique and electrochemical methods. It crystallizes in the tetragonal crystal systems with space group I4(1)cd and Z=8. The copper(II)
K Sugibayashi et al.
Journal of controlled release : official journal of the Controlled Release Society, 62(1-2), 201-208 (1999-10-16)
An in vitro permeation study of ethyl nicotinate (EN) was carried out using excised hairless rat skin, and simultaneous skin transport and metabolism of the drug were kinetically followed. Fairly good steady-state fluxes of EN and its metabolite nicotinic acid
Tetsuya Hasegawa et al.
Biological & pharmaceutical bulletin, 31(1), 85-89 (2008-01-08)
The skin disposition and metabolism of topically applied ethyl nicotinate (EN) were evaluated in dual agar gel disc-inserted hairless rats, which have two agar gel discs subcutaneously inserted into the abdominal region as drug receptors, and a topical formulation containing
T Ngawhirunpat et al.
Biological & pharmaceutical bulletin, 24(4), 414-417 (2001-04-18)
The age-dependent characteristics of transdermal permeation of ethyl nicotinate (EN) and its metabolism to nicotinic acid (NA) were examined in rats at the ages of a fetus at 21 d, 3, 10, 50, 270 and 360 d. Skin transport in
K Sugibayashi et al.
Pharmaceutical research, 13(6), 855-860 (1996-06-01)
Simultaneous skin transport and metabolism of ethyl nicotinate (EN), a model drug, were measured and theoretically analyzed. Several permeation studies of EN or its metabolite nicotinic acid (NA) were done on full-thickness skin or stripped skin with and without an

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service