Skip to Content
Merck
All Photos(3)

Documents

E36652

Sigma-Aldrich

2-Ethyl-4-methylimidazole

95%

Synonym(s):

EMI-2,4

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H10N2
CAS Number:
Molecular Weight:
110.16
Beilstein:
1711
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

refractive index

n20/D 1.5 (lit.)

bp

292-295 °C (lit.)

mp

47-54 °C (lit.)

density

0.975 g/mL at 25 °C (lit.)

SMILES string

CCc1nc(C)c[nH]1

InChI

1S/C6H10N2/c1-3-6-7-4-5(2)8-6/h4H,3H2,1-2H3,(H,7,8)

InChI key

ULKLGIFJWFIQFF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Ethyl-4-methylimidazole can be used as a:
  • cross-linker for epoxy resins to synthesize network polymers with improved thermal resistance and physical properties.
  • curing agent for polysiloxane episulfide resin (PSER).
  • catalyst to promote curing of blends of hyperbranched aromatic epoxy polymer and bisphenol F diglycidyl ether.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

278.6 °F - closed cup

Flash Point(C)

137 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Co-cross-linking of bio-based multi-functional epoxide and bisphenol A diglycidyl ether with 2-ethyl-4-methylimidazole.
Morinaga H and Sakamoto M
Tetrahedron Letters, 59(43), 3889-3891 (2018)
Hyperbranched aromatic epoxies in the design of adhesive materials.
Emrick T, et al.
Polymer Bull., 45(1), 1-7 (2000)
Yuezhan Feng et al.
Journal of hazardous materials, 346, 140-151 (2017-12-20)
Phosphorus and/or nitrogen doping is an effective method of improving the physical and chemical properties of reduced graphene oxide (rGO). In this work, phosphorus and nitrogen co-doped rGO (PN-rGO), synthesized using a scalable hydrothermal and microwave process, was used as
Van-Dung Mai et al.
Polymers, 11(2) (2019-04-10)
A bio-derived dihydroxylimine hardener, Van2HMDA, for the curing of epoxy resin was prepared from vanillin (Van) and hexamethylene-1,6-diamine (HMDA) by Schiff base formation. The epoxy resin of diglycidyl ether of bisphenol A was cured with Van2HMDA in the presence of
Cheng Hao et al.
ChemSusChem, 12(5), 1049-1058 (2018-12-12)
Conventional thermoset coatings cannot be easily repaired and removed owing to their highly crosslinked structure. The investigation of repairable or removable coatings has been receiving extensive attention, but few reported coatings possess both features. In this work, a repairable and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service