Skip to Content
Merck
All Photos(3)

Documents

E16600

Sigma-Aldrich

9-Ethylcarbazole

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H13N
CAS Number:
Molecular Weight:
195.26
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

68-70 °C (lit.)

SMILES string

CCn1c2ccccc2c3ccccc13

InChI

1S/C14H13N/c1-2-15-13-9-5-3-7-11(13)12-8-4-6-10-14(12)15/h3-10H,2H2,1H3

InChI key

PLAZXGNBGZYJSA-UHFFFAOYSA-N

Features and Benefits

Can be used as a plasticizer in guest-host polymers to yield highly efficient photorefractive polymer composites.

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Chimento et al.
Journal of wound care, 26(Sup9), S4-S8 (2017-09-08)
Osteopontin (OPN) is abundantly expressed during tissue repair, acting as a powerful chemokine that recruits inflammatory cells such as neutrophils, macrophages, and Langerhans cells. The role of OPN in chronic wounds has not been explored. In this study, we assess
Denzil Britto Christopher Leslee et al.
Luminescence : the journal of biological and chemical luminescence, 32(7), 1354-1360 (2017-06-21)
A new carbazole-azine based fluorescent sensor was synthesized and characterized. The selectivity of the sensor for Cu
Kim van der Weerd et al.
Thyroid : official journal of the American Thyroid Association, 24(6), 1051-1061 (2014-03-19)
Using gene expression profiling, we detected differential thyrotropin receptor (TSH-R) expression during human T-cell development in the thymus. This expression pattern indicated a potential role for the TSH-R within the thymus, independent of its function in the thyroid gland. Here
Guanglin Cui et al.
Oncotarget, 8(28), 45311-45322 (2017-05-10)
Considerable evidence supports the idea that stem-like cells may play an essential role during the development of colorectal cancer (CRC). To accomplish this aim, we use immunohistochemistry (IHC) and double IHC with different potential stem-like markers, anti-musashi (Msi), anti-CD133, anti-
Keith R Spencer et al.
Brain pathology (Zurich, Switzerland), 30(6), 1028-1040 (2020-07-08)
Amyotrophic lateral sclerosis (ALS) is a progressive neurodegenerative disorder affecting both the upper and lower motor neurons. Although ALS typically leads to death within 3 to 5 years after initial symptom onset, approximately 10% of patients with ALS live more than

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service