Skip to Content
Merck
All Photos(2)

Key Documents

E10505

Sigma-Aldrich

Ethyl 3-aminobenzoate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC6H4CO2C2H5
CAS Number:
Molecular Weight:
165.19
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.560 (lit.)

bp

172-175 °C/13 mmHg (lit.)

density

1.107 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)c1cccc(N)c1

InChI

1S/C9H11NO2/c1-2-12-9(11)7-4-3-5-8(10)6-7/h3-6H,2,10H2,1H3

InChI key

ZMCBYSBVJIMENC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marta Lozano-Ortega et al.
PLoS biology, 16(7), e2004404-e2004404 (2018-07-20)
Directional mechanoreception by hair cells is transmitted to the brain via afferent neurons to enable postural control and rheotaxis. Neuronal tuning to individual directions of mechanical flow occurs when each peripheral axon selectively synapses with multiple hair cells of identical
S Bartkova et al.
Journal of fish diseases, 40(1), 73-82 (2016-06-17)
Recent development of imaging tools has facilitated studies of pathogen infections in vivo in real time. This trend can be exemplified by advances in bioluminescence imaging (BLI), an approach that helps to visualize dissemination of pathogens within the same animal over
Xiaolin Zhang et al.
International journal of nanomedicine, 13, 5377-5394 (2018-09-27)
Infections such as biomaterial-associated infection and osteomyelitis are often associated with intracellular survival of bacteria (eg, Staphylococcus aureus). Treatment of these infections remains a major challenge due to the low intracellular efficacy of many antibiotics. Therefore, local delivery systems are
Lucas A Zena et al.
Scientific reports, 7(1), 16119-16119 (2017-11-25)
Beat-to-beat variation in heart rate (f
Koh Araishi et al.
Peptides, 119, 170118-170118 (2019-07-08)
Arginine vasotocin (AVT) is known as a neurohypophyseal hormone that regulates water- and mineral-balance in non-mammalian vertebrates. Recent studies revealed that AVT also exerts central effects on behavior. The goldfish has several merits for evaluation of behavioral changes. However, there

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service