DWK241702603
DURAN® reaction flask with ground joint
glass flask, flask capacity (100 mL), single neck, round bottom, neck joint: ST/NS 24/29
Synonym(s):
241702603
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
Recommended Products
material
glass flask
round bottom
Joint
joint: ST/NS neck24/29
feature
lid
packaging
case of 10 ea
manufacturer/tradename
DURAN® 241702603
flask capacity
(100 mL)
Looking for similar products? Visit Product Comparison Guide
General description
Tight closure due to standard ground joint. The geometry permits very uniform heating. Closable with glass and plastic stopper
Features and Benefits
- Complies with DIN ISO 4797
- Manufactured from 3.3 expansion borosilicate glass
- Tight closure due to standard interchangeable ground neck joint
- The geometry permits very uniform heating
- Closable with glass and plastic stopper
- Fired-on, highly durable white print
Legal Information
DURAN is a registered trademark of DWK Life Sciences
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service