Skip to Content
Merck
All Photos(3)

Documents

D60400

Sigma-Aldrich

2,3-Dichloro-5,6-dicyano-p-benzoquinone

98%

Synonym(s):

4,5-Dichloro-3,6-dioxo-1,4-cyclohexadiene-1,2-dicarbonitrile, DDQ

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8Cl2N2O2
CAS Number:
Molecular Weight:
227.00
Beilstein:
747939
EC Number:
MDL number:
UNSPSC Code:
12352117
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

powder

reaction suitability

reagent type: oxidant

mp

210-215 °C (dec.) (lit.)

storage temp.

2-8°C

SMILES string

ClC1=C(Cl)C(=O)C(C#N)=C(C#N)C1=O

InChI

1S/C8Cl2N2O2/c9-5-6(10)8(14)4(2-12)3(1-11)7(5)13

InChI key

HZNVUJQVZSTENZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2,3-Dichloro-5,6-dicyano-p-benzoquinone (DDQ) can be used as:
  • A deprotecting reagent for a variety of compounds, such as thioacetals, acetals, and ketals.
  • An electron-transfer reagent for the synthesis of quinolines from imines and alkynes or alkenes.
  • An effective reagent for the benzylic and allylic C−H functionalization.
  • An oxidizing agent for the synthesis of functionalized furans and benzofurans.
  • A reagent with Ph3P in an efficient synthesis of 1,2-benzisoxazoles.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Supplementary Hazards

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

DDQ-mediated formation of carbon? carbon bonds: Oxidation of imines
Bortolotti B, et al.
Tetrahedron, 49, 10157-10157 (1993)
Ayse Kocabalkanli et al.
Farmaco (Societa chimica italiana : 1989), 57(12), 993-997 (2003-02-05)
Nucleosides have been of great interest since their strong antiviral activities were discovered. 1,3-Oxathiolane ring system has been known for many years, but it is in recent years that the ring has been used as the sugar ring in nucleoside
Predictive model for oxidative C-H bond functionalization reactivity with 2, 3-dichloro-5, 6-dicyano-1, 4-benzoquinone.
Morales-Rivera CA, et al.
Journal of the American Chemical Society, 139(49), 17935-17944 (2017)
Mild and efficient photochemical and thermal deprotection of thioacetals and ketals using DDQ
Mathew L and Sankararaman S
The Journal of Organic Chemistry, 58, 7576-7576 (1993)
Deprotection of benzyl ethers using 2, 3-dichloro-5, 6-dicyano-p-benzoquinone (DDQ) under photoirradiation.
Rahim MA, et al.
Tetrahedron Letters, 46(43), 7307-7309 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service