Skip to Content
Merck
All Photos(2)

Key Documents

D56829

Sigma-Aldrich

1,4-Dichlorobenzene

≥99%

Synonym(s):

1,4-Dichlorobenzene, Di-chloricide, Dichlorocide, PDB, Paradichlorobenzene, p-Chlorophenyl chloride, p-Dichlorobenzene, para-Dichlorobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H4Cl2
CAS Number:
Molecular Weight:
147.00
Beilstein:
1680023
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay:
≥99%
bp:
173 °C (lit.)
vapor pressure:
1.03 mmHg ( 25 °C)

vapor density

5.07 (vs air)

Quality Level

vapor pressure

1.03 mmHg ( 25 °C)

Assay

≥99%

form

crystalline solid

bp

173 °C (lit.)

mp

52-54 °C (lit.)

density

1.241 g/mL at 25 °C (lit.)

SMILES string

Clc1ccc(Cl)cc1

InChI

1S/C6H4Cl2/c7-5-1-2-6(8)4-3-5/h1-4H

InChI key

OCJBOOLMMGQPQU-UHFFFAOYSA-N

Gene Information

human ... CYP2A6(1548)
mouse ... Cyp2a5(13087)

Looking for similar products? Visit Product Comparison Guide

Application

1,4-Dichlorobenzene can be used as a reactant:
  • In the synthesis of nitro-aromatics via electrophilic nitration.
  • To synthesize polyarylbenzenes through C-C cross coupling with arylboronic acids using imidazolium salt as a catalyst.
  • In the Pd-catalyzed Mizoroki-Heck coupling reaction with alkenes.

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ilknur Hatice Eryilmaz et al.
Scientific reports, 7(1), 10526-10526 (2017-09-07)
Self-assembly methods allow to obtain ordered patterns on surfaces with exquisite precision, but often lack in effectiveness over large areas. Here we report on the realization of hierarchically ordered polymethylmethacrylate (PMMA) nanofibres and nanodots over large areas from solution via
Osamu Takahashi et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 49(1), 49-56 (2010-10-12)
Rats and mice (8 animals per species per group) were injected subcutaneously or intraperitoneally with paradichlorobenzene (PDCB) at doses of 0, 100, 200 and 400 mg/kg/day, 4-5 days a week, for 8 weeks (for rats) and either 2 or 6
Michael C McCarthy et al.
Environmental health perspectives, 117(5), 790-796 (2009-05-30)
Ambient measurements of hazardous air pollutants (air toxics) have been used to validate model-predicted concentrations of air toxics but have not been used to perform risk screening at the national level. We used ambient concentrations of routinely measured air toxics
Diana E Hun et al.
Environmental health perspectives, 117(12), 1925-1931 (2010-01-06)
Hispanics are the fastest growing minority group in the United States; however, minimal information is available on their cancer risks from exposures to hazardous air pollutants (HAPs) and how these risks compare to risks to non-Hispanic whites. We estimated the
Minjin Lee et al.
International journal of environmental research and public health, 15(4) (2018-04-14)
The inhalation of a water aerosol from a humidifier containing disinfectants has led to serious lung injuries in Korea. To promote the safe use of products, the Korean government enacted regulations on the chemicals in various consumer products that could

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service