Skip to Content
Merck
All Photos(1)

Key Documents

D49598

Sigma-Aldrich

N,N′-Dibutylthiourea

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3NHCSNH(CH2)3CH3
CAS Number:
Molecular Weight:
188.33
Beilstein:
507434
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

powder

mp

63-65 °C (lit.)

SMILES string

CCCCNC(=S)NCCCC

InChI

1S/C9H20N2S/c1-3-5-7-10-9(12)11-8-6-4-2/h3-8H2,1-2H3,(H2,10,11,12)

InChI key

KFFQABQEJATQAT-UHFFFAOYSA-N

Gene Information

human ... EPHX2(2053)
mouse ... Ephx2(13850)

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Occupational airborne allergic contact dermatitis from dibutylthiourea.
L Kanerva et al.
Contact dermatitis, 38(6), 347-348 (1998-08-01)
Erin M Warshaw et al.
Dermatitis : contact, atopic, occupational, drug, 19(4), 190-201 (2008-08-05)
Allergic contact dermatitis from thioureas is uncommon but may result from exposure to rubber, especially neoprene. To (1) describe the population with positive patch-test reactions to mixed dialkyl thioureas (MDTU) (ie, diethylthiourea and dibutylthiourea); (2) determine clinical and occupational relevance
Allergic contact dermatitis from dibutylthiourea in a wet suit.
Venkat S Gudi et al.
Dermatitis : contact, atopic, occupational, drug, 15(1), 55-56 (2004-12-03)
Contact dermatitis to dibutylthiourea in swimming goggles.
A Alomar et al.
Contact dermatitis, 13(5), 348-349 (1985-11-01)
Contact dermatitis due to occupational dibutylthiourea exposure: a case of phonecard dermatitis.
P Schmid-Grendelmeier et al.
Contact dermatitis, 32(5), 308-309 (1995-05-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service