Skip to Content
Merck
All Photos(1)

Key Documents

D165603

Sigma-Aldrich

1,2-Dimethylindole

99%

Synonym(s):

NSC 62087

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H11N
CAS Number:
Molecular Weight:
145.20
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

crystals

mp

55-58 °C (lit.)

SMILES string

Cc1cc2ccccc2n1C

InChI

1S/C10H11N/c1-8-7-9-5-3-4-6-10(9)11(8)2/h3-7H,1-2H3

InChI key

BJMUOUXGBFNLSN-UHFFFAOYSA-N

Application

  • Building block for preparation of 3-functionalized indoles
  • Building block for preparation carbazoles
  • Building block for preparation of cyanoindoles
  • Reactant for preparation of trifluoromethylindoles
  • Reactant for preparation of potent antihyperlipidemic agents
  • Building block in iso-Nazarov or Nazarov cyclizations
  • Reactant for preparation of GSK-3ß inhibitors
  • Building block for preparation of carboxyindoles
  • Reactant for preparation of plant growth promoters
  • Reactant for preparation of tryptophan derivatives

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service