Skip to Content
Merck
All Photos(1)

Key Documents

CDS000697

Sigma-Aldrich

Sodium hexadecyl sulfate

AldrichCPR

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H33NaO4S
Molecular Weight:
344.49
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

solid

SMILES string

[Na+].CCCCCCCCCCCCCCCCOS([O-])(=O)=O

InChI

1S/C16H34O4S.Na/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-20-21(17,18)19;/h2-16H2,1H3,(H,17,18,19);/q;+1/p-1

InChI key

GGHPAKFFUZUEKL-UHFFFAOYSA-M

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qian Guan et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 1152, 122230-122230 (2020-06-26)
Capillary gel electrophoresis (CGE) using sodium dodecyl sulfate (CGE-SDS or CE-SDS) is commonly used in the biotechnology industry to assess the purity of a complex therapeutic during manufacturing process optimization and also for commercial release and stability testing. However, for
Concepción Abril et al.
Chemosphere, 268, 128823-128823 (2020-11-09)
The uptake, bioconcentration and translocation of 22 endocrine disrupting compounds (six perflurocarboxylic acids (PFAAs), perfluorooctanoic sulfonic acid, four anionic surfactants (alkylsulfates (ASC) from C12 to C16), bisphenol A (BPA), four preservatives (parabens), two biocides (triclosan (TCS) and triclocarban (TCB)) and
Xuecong Pan et al.
Langmuir : the ACS journal of surfaces and colloids, 34(18), 5273-5278 (2018-04-20)
Cooperative effects of a series of equimolar binary zwitterionic-ionic surfactant mixtures on the interfacial water structure at the air-water interfaces have been studied by sum frequency generation vibrational spectroscopy (SFG-VS). For zwitterionic surfactant palmityl sulfobetaine (SNC16), anionic surfactant sodium hexadecyl
Concepción Abril et al.
Analytical and bioanalytical chemistry, 410(21), 5155-5163 (2018-06-28)
An analytical method based on ultrasound-assisted extraction and dispersive liquid-liquid microextraction (DLLME) clean-up has been developed and validated for the determination of 31 emerging pollutants in root vegetables. The target compounds were four preservatives, six perfluoroalkyl compounds, six UV filters
Qiang Zhao et al.
Talanta, 151, 209-216 (2016-03-08)
In this study, surface-functionalized nitrogen-doped graphene was prepared with 1, 3, 6, 8-pyrenetetrasulfonic acid tetrasodium salt in a simple way. The characteristics of the material were demonstrated by UV-vis absorption spectra, X-ray photoelectron spectroscopy, and electrochemical experiments. Then, the glassy

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service