Skip to Content
Merck
All Photos(3)

Key Documents

C82604

Sigma-Aldrich

Citraconic acid

98%

Synonym(s):

Methylmaleic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C(CO2H)=CHCO2H
CAS Number:
Molecular Weight:
130.10
Beilstein:
1722679
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

crystals

mp

88-94 °C (lit.)

SMILES string

C\C(=C\C(O)=O)C(O)=O

InChI

1S/C5H6O4/c1-3(5(8)9)2-4(6)7/h2H,1H3,(H,6,7)(H,8,9)/b3-2-

InChI key

HNEGQIOMVPPMNR-IHWYPQMZSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nikolai P Bityutskii et al.
Journal of plant physiology, 218, 100-108 (2017-08-19)
Aluminium (Al) is one of the major stressors for plants in acidic soils, negatively affecting plant growth and nutrient balances. Significant efforts have been undertaken to understand mechanisms of Al tolerance in plants. However, little is known of the relevance
Ananya Sen et al.
Journal of bacteriology, 202(18) (2020-07-01)
Hydrogen peroxide (H2O2) is formed in natural environments by both biotic and abiotic processes. It easily enters the cytoplasms of microorganisms, where it can disrupt growth by inactivating iron-dependent enzymes. It also reacts with the intracellular iron pool, generating hydroxyl
Nicolle H R Litjens et al.
European journal of immunology, 34(2), 565-575 (2004-02-10)
Psoriasis vulgaris, a type-1 cytokine-mediated chronic skin disease, can be treated successfully with fumaric acid esters (FAE). Beneficial effects of this medication coincided with decreased production of IFN-gamma. Since dendritic cells (DC) regulate the differentiation of T helper (Th) cells
Darius Moharregh-Khiabani et al.
PloS one, 5(7), e11769-e11769 (2010-07-30)
Fumaric acid esters (FAE) are a group of compounds which are currently under investigation as an oral treatment for relapsing-remitting multiple sclerosis. One of the suggested modes of action is the potential of FAE to exert a neuroprotective effect. We
Maria Khomyakova et al.
Science (New York, N.Y.), 331(6015), 334-337 (2011-01-22)
Access to novel ecological niches often requires adaptation of metabolic pathways to cope with new environments. For conversion to cellular building blocks, many substrates enter central carbon metabolism via acetyl-coenzyme A (acetyl-CoA). Until now, only two such pathways have been

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service