Skip to Content
Merck
All Photos(1)

Key Documents

C102202

Sigma-Aldrich

Cyclohexanone oxime

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H10(=NOH)
CAS Number:
Molecular Weight:
113.16
Beilstein:
1616769
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

crystals

bp

206-210 °C (lit.)

mp

86-89 °C (lit.)

SMILES string

O\N=C1/CCCCC1

InChI

1S/C6H11NO/c8-7-6-4-2-1-3-5-6/h8H,1-5H2

InChI key

VEZUQRBDRNJBJY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shelli J Schomaker et al.
Toxicology and applied pharmacology, 185(1), 48-54 (2002-12-04)
The purpose of these studies was to evaluate bone marrow from male CD rats following exposure to known hematotoxins using flow cytometry (FC) and a monoclonal antibody to the cell surface antigen CD71. Rats were treated with either CHO (300
S C Gad et al.
Fundamental and applied toxicology : official journal of the Society of Toxicology, 5(1), 128-136 (1985-02-01)
Dermal exposure of rabbits to cyclohexanone oxime (CHO) for 24 hr at 0, 0.8, 2, and 5 g/kg caused dose-related reticulocytosis on the day after dosing as well as a decrease in hemoglobin in the 5-g/kg females 7 days postdosing.
Wen-Cui Li et al.
Journal of the American Chemical Society, 127(36), 12595-12600 (2005-09-08)
In this study, we present a synthetic pathway for the fabrication of self-supporting zeolite monoliths consisting of crystallized nanoparticles. A resorcinol-formaldehyde-based organic aerogel is used as a template, and silicalite-1 is used as the zeolite example. The silicalite-1 monoliths obtained
Raphaël Turgis et al.
ChemSusChem, 3(12), 1403-1408 (2010-12-01)
Brønsted-acidic ionic liquids that bear a sulfonic acid group, known as Forbes acids, show a good catalytic activity for the Beckmann rearrangement, used to prepare ε-caprolactam, which is a precursor of Nylon 6. The activity essentially stems from the acidity of
M J Derelanko et al.
Fundamental and applied toxicology : official journal of the Society of Toxicology, 5(1), 117-127 (1985-02-01)
Cyclohexanone oxime (CHO) was given po to male and female Fischer 344 rats at dose levels of 10, 25, 75, 150, and 300 mg/kg, five times a week for a period of 2 weeks. Control animals received distilled water. All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service