C101400
1,2-Cyclohexanedione
97%
Synonym(s):
1,2-Dioxocyclohexane, Cyclohexan-1,2-dione
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Linear Formula:
C6H8(=O)2
CAS Number:
Molecular Weight:
112.13
Beilstein:
507419
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
97%
form
solid
bp
193-195 °C (lit.)
mp
34-38 °C (lit.)
storage temp.
2-8°C
SMILES string
O=C1CCCCC1=O
InChI
1S/C6H8O2/c7-5-3-1-2-4-6(5)8/h1-4H2
InChI key
OILAIQUEIWYQPH-UHFFFAOYSA-N
Gene Information
human ... ACHE(43) , BCHE(590) , CES1(1066)
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Specific reagent for arginine residues.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
No data available
Flash Point(C)
No data available
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
D Scott Wilbur et al.
Bioconjugate chemistry, 13(3), 611-620 (2002-05-16)
Recombinant streptavidin (rSAv) is of interest as a carrier of alpha-emitting radionuclides in pretargeting protocols for cancer therapy. Due to the inherently high kidney localization of rSAv, modification of this protein is required before it can be useful in pretargeting.
S Adak et al.
The Biochemical journal, 314 ( Pt 3), 985-991 (1996-03-15)
The plausible role of arginine and tyrosine residues at the active side of horseradish peroxidase (HRP) in aromatic donor (guaiacol) oxidation was probed by chemical modification followed by characterization of the modified enzyme. The arginine-specific reagents phenylglyoxal (PGO), 2,3-butanedione and
C Cervenanský et al.
European journal of biochemistry, 229(1), 270-275 (1995-04-01)
The West African green mamba, Dendroaspis angusticeps, has two toxins, fasciculins, that are non-competitive inhibitors of acetylcholinesterase. Arginine residues of fasciculin 2 were modified with 1,2-cyclohexanedione. Two of these residues, Arg24 and Arg37, reacted very slowly or not at all.
Andres De la Rossa et al.
Nature neuroscience, 16(2), 193-200 (2013-01-08)
The molecular mechanisms that control how progenitors generate distinct subtypes of neurons, and how undifferentiated neurons acquire their specific identity during corticogenesis, are increasingly understood. However, whether postmitotic neurons can change their identity at late stages of differentiation remains unknown.
Bellamkonda Ramakrishna et al.
International journal of biological macromolecules, 115, 1225-1232 (2018-05-05)
The recombinant C-terminal domain of chitinase C of Chitinophaga pinensis (CpChiC-GH18C) exhibits the highest activity at pH 6.0 and 35 °C, with a Km of 76.13 (mg-1 ml), a kcat of 10.16 (s-1), and a kcat/Km of 0.133 (mg-1 ml s-1) on colloidal chitin. Analysis
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service