Skip to Content
Merck
All Photos(1)

Key Documents

BR781412

BRAND® 24-well PCR plate

no skirt, white

Synonym(s):

PCR, PCR Plates, Plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106300
NACRES:
NB.22
material:
polypropylene
white
white wells
size:
24 well
binding type:
non-treated surface
feature:
skirt: no
no skirt

material

polypropylene
white
white wells

description

D-Nase/R-Nase, DNA-free, suitable for qPCR

feature

skirt: no
no skirt

packaging

pack of 40 ea

manufacturer/tradename

BRAND 781412

technique(s)

PCR: suitable

size

24 well

well size

0.2 mL

working volume

200 μm

binding type

non-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

These plates reduce material costs and allow work on a compact PCR plate even with small sample throughputs. The ultra thin-walled PCR plate design facilitates constant, rapid and precise heat transfer. The smooth vessel interior minimizes the binding of enzymes and nucleic acid to the walls. The raised lips of the wells protect against cross-contamination, allowing a reliable seal with the sealing mat tailored to the plates.

Features and Benefits

  • Fits all common thermal cycler
  • Raised well edges protect against cross-contamination
  • Rapid heat transfer through thin vessel walls

Legal Information

BRAND is a registered trademark of BRAND GMBH + CO KG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service