Merck
BR458024
BRAND® filter crucible
borosilicate glass, capacity 30 mL, 1 D 4
Synonym(s):
Filter, Filter crucible
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NB.43
UNSPSC Code:
41122200
Manufacturer/tradename:
BRAND 458024
Capacity:
30 mL
Material:
borosilicate glass
Pore size:
10-16 μm
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
material
borosilicate glass
manufacturer/tradename
BRAND 458024
capacity
30 mL
pore size
10-16 μm
General description
The BRAND® Filter Crucible made from Boro 3.3 glass is an essential tool for efficient filtration and separation processes in laboratory applications.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Still not finding the right product?
Explore all of our products under BRAND® filter crucible
Choose from one of the most recent versions:
Certificates of Analysis (COA)