Skip to Content
Merck
All Photos(2)

Key Documents

B93989

Sigma-Aldrich

Dibutyl disulfide

97%

Synonym(s):

Butyl disulfide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3SS(CH2)3CH3
CAS Number:
Molecular Weight:
178.36
Beilstein:
1735184
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
liquid
Assay:
97%

vapor pressure

52 mmHg ( 37.7 °C)

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.492 (lit.)

bp

229-233 °C (lit.)

density

0.938 g/mL at 25 °C (lit.)

SMILES string

CCCCSSCCCC

InChI

1S/C8H18S2/c1-3-5-7-9-10-8-6-4-2/h3-8H2,1-2H3

InChI key

CUDSBWGCGSUXDB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

199.4 °F - closed cup

Flash Point(C)

93 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Lóránd Kiss et al.
Toxicological sciences : an official journal of the Society of Toxicology, 160(2), 398-407 (2017-10-04)
Recent in vitro and in vivo studies highlight the strong potential of dimethyl trisulfide (DMTS) as an antidote for cyanide (CN) intoxication. Due to its high oxygen demand, the brain is one of the main target organs of CN. The
K M Abdo et al.
Fundamental and applied toxicology : official journal of the Society of Toxicology, 3(2), 69-74 (1983-03-01)
n-Butyl mercaptan (nBM) is a breakdown product of S,S,S,-tri-n-butyl phosphorotrithioate (DEF) and S,S,S-tri-n-butyl phosphorotrithioite (merphos) in hens and in the environment. n-Butyl disulfide (nBD) is an oxidation product of nBM. A single 500 mg/kg dose of nBM and nBD was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service