All Photos(3)
About This Item
Linear Formula:
BrC10H6OH
CAS Number:
Molecular Weight:
223.07
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
97%
form
powder
mp
122-124 °C (lit.)
SMILES string
Oc1ccc2cc(Br)ccc2c1
InChI
1S/C10H7BrO/c11-9-3-1-8-6-10(12)4-2-7(8)5-9/h1-6,12H
InChI key
YLDFTMJPQJXGSS-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
K Nakashima et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 58(1), 123-128 (2002-01-26)
Room temperature phosphorescence (RTP) of 6-bromo-2-naphthol has been investigated in aqueous micellar solutions of poly(ethylene oxide)-poly(propylene oxide)-poly(ethylene oxide) block copolymers as well as in their mixed aggregates with sodium dodecyl sulfate. RTP of the phosphorophor was enhanced to some extent
F Tanaka et al.
Bioorganic & medicinal chemistry letters, 11(22), 2983-2986 (2001-10-26)
We developed a visible detection system for antibody-catalyzed retro-aldol-retro-Michael reactions. Aldolase antibody 38C2 catalyzed the reaction of substrate 1 to provide 6-bromo-2-napthol that forms a visible colored azo dye with diazonium salts. This system has potential for the screening of
Chaoren Shen et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 23(54), 13369-13378 (2017-06-27)
The palladium-catalyzed chemoselective carbonylation of bromoaryl triflates is reported. The selective C-Br bond versus C-OTf (OTf=triflate) bond functionalization can be remarkably tuned by the combination of the ligand [4,5-bis(diphenylphosphino)-9,9-dimethylxanthene (Xantphos) vs. 1,1'-bis(diphenylphosphino)ferrocene (DPPF)] and the solvent (toluene vs. DMSO). The
A specific stain for alpha-glucosidases in isoelectric focusing gels.
L L Spielman et al.
Analytical biochemistry, 120(1), 66-70 (1982-02-01)
Win-Long Chia et al.
International journal of molecular sciences, 16(4), 7520-7534 (2015-04-08)
A new series of 2-(6-alkoxynaphthalen-2-yl)-6-methoxyquinolines (nO-NpQOMe, n = 3-8) liquid crystal compounds, a linear molecular structure with two kinks, were synthesized using a short two-step reaction with overall yields between 43% and 58%. Spectral analyses were in accord with the
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service