Skip to Content
Merck
All Photos(1)

Documents

B68240

Sigma-Aldrich

1-Bromohexane

98%

Synonym(s):

Hexyl bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)5Br
CAS Number:
Molecular Weight:
165.07
Beilstein:
1731290
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

5.7 (vs air)

Quality Level

vapor pressure

<10 mmHg ( 20 °C)

Assay

98%

form

liquid

refractive index

n20/D 1.448 (lit.)

bp

154-158 °C (lit.)

mp

−85 °C (lit.)

density

1.176 g/mL at 25 °C (lit.)

SMILES string

CCCCCCBr

InChI

1S/C6H13Br/c1-2-3-4-5-6-7/h2-6H2,1H3

InChI key

MNDIARAMWBIKFW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

134.6 °F - closed cup

Flash Point(C)

57 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sitaraman Krishnan et al.
Langmuir : the ACS journal of surfaces and colloids, 22(26), 11255-11266 (2006-12-13)
Polystyrene-b-poly(4-vinylpyridine) copolymers were quaternized with 1-bromohexane and 6-perfluorooctyl-1-bromohexane. Surfaces prepared from these polymers were characterized by contact angle measurements, near-edge X-ray absorption fine structure spectroscopy and X-ray photoelectron spectroscopy. The fluorinated pyridinium surfaces showed enhanced antibacterial activity compared to their
R J Pieters et al.
Bioorganic & medicinal chemistry letters, 9(2), 161-166 (1999-02-18)
Bifunctional molecules containing both a biotin and a substrate unit have been designed and synthesized for phage display screening of mutant libraries of haloalkane dehalogenase enzymes. The molecules were assembled using a convergent modular synthetic strategy. One molecule was synthesized
Halina R Novak et al.
FEBS letters, 588(9), 1616-1622 (2014-03-13)
A putative haloalkane dehalogenase has been identified in a marine Rhodobacteraceae and subsequently cloned and over-expressed in Escherichia coli. The enzyme has highest activity towards the substrates 1,6-dichlorohexane, 1-bromooctane, 1,3-dibromopropane and 1-bromohexane. The crystal structures of the enzyme in the
Mitsuru Abe et al.
Physical chemistry chemical physics : PCCP, 17(48), 32276-32282 (2015-11-20)
We have synthesised novel ionic liquids (ILs) to show both cellulose dissolution ability and LCST-type phase transition after mixing with water. To realise both polar and hydrophobic properties, tetraalkylphosphonium cations and a series of carboxylate anions were employed to assume
Sedigheh Kamran et al.
Amino acids, 47(12), 2483-2493 (2015-07-08)
In this paper, nanoparticles of Fe3O4 as well as their modified forms with different ionic liquids (IL-Fe3O4) were prepared and used for adsorption of lysozyme. The mean size and the surface morphology of the nanoparticles were characterized by TEM, XRD

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service