Skip to Content
Merck
All Photos(1)

Key Documents

ALD01654

Sigma-Aldrich

3-((2-methoxypyridin-3-yl) amino)-3-methylbutan-1-ol

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

form

liquid

storage temp.

2-8°C

SMILES string

CC(CCO)(C)NC1=CC=CN=C1OC

InChI

1S/C11H18N2O2/c1-11(2,6-8-14)13-9-5-4-7-12-10(9)15-3/h4-5,7,13-14H,6,8H2,1-3H3

InChI key

DORIBENRPNIVDI-UHFFFAOYSA-N

Application

Traditionally challenging to access, this hindered amine provides a chemically differentiated building block for organic synthesis and medicinal chemistry, specifically for preparation of drug candidates containing hindered amine motifs. Additionally, this and over 80 other sterically hindered secondary amines in the Sigma-Aldrich portfolio provide structurally distinct screening compounds. Screen several through Aldrich Market Select. This hindered amine was accessed via a recent publication reported by Baran and coworkers using an innovative hydroamination method.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Baran Group offers zinc-based reagents for versatile transformations in organic synthesis.

Baran Group offers zinc-based reagents for versatile transformations in organic synthesis.

Baran Group offers zinc-based reagents for versatile transformations in organic synthesis.

Baran Group offers zinc-based reagents for versatile transformations in organic synthesis.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service