Skip to Content
Merck
All Photos(1)

Key Documents

ALD00526

Sigma-Aldrich

Serine Hydrolase Inhibitor-9

Synonym(s):

tert-Butyl (2-(3-(2-fluorophenyl)-1-methylureido)ethyl)(methyl)carbamate, SHI-9

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H24FN3O3
CAS Number:
Molecular Weight:
325.38
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

form

powder

Quality Level

SMILES string

O=C(N(CCN(C(OC(C)(C)C)=O)C)C)NC1=C(F)C=CC=C1

InChI

1S/C16H24FN3O3/c1-16(2,3)23-15(22)20(5)11-10-19(4)14(21)18-13-9-7-6-8-12(13)17/h6-9H,10-11H2,1-5H3,(H,18,21)

InChI key

OOSVRVOVTXLHKA-UHFFFAOYSA-N

Application

Serine hydrolase inhibitors are small molecule fragments that react in vitro with the serine hydrolase class of proteins. They are amenable to further modification to create novel inhibitors of this class of protein.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katerina Otrubova et al.
Bioorganic & medicinal chemistry letters, 24(16), 3807-3813 (2014-07-20)
Two libraries of modestly reactive ureas containing either electron-deficient acyl anilines or acyl pyrazoles were prepared and are reported as screening libraries for candidate serine hydrolase inhibitors. Within each library is a small but powerful subset of compounds that serve

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service