Skip to Content
Merck
All Photos(2)

Key Documents

A86805

Sigma-Aldrich

Aniline-2-sulfonic acid

95%

Synonym(s):

Orthanilic acid, 2-Aminobenzenesulfonic acid, o-Sulfanilic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC6H4SO3H
CAS Number:
Molecular Weight:
173.19
Beilstein:
1309204
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

form

solid

SMILES string

Nc1ccccc1S(O)(=O)=O

InChI

1S/C6H7NO3S/c7-5-3-1-2-4-6(5)11(8,9)10/h1-4H,7H2,(H,8,9,10)

InChI key

ZMCHBSMFKQYNKA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

J Mampel et al.
Microbiology (Reading, England), 145 ( Pt 11), 3255-3264 (1999-12-10)
Growth of Alcaligenes sp. strain O-1 with 2-aminobenzenesulfonate (ABS; orthanilate) as sole source of carbon and energy requires expression of the soluble, multicomponent 2-aminobenzenesulfonate 2,3-dioxygenase system (deaminating) (ABSDOS) which is plasmid-encoded. ABSDOS was separated by anion-exchange chromatography to yield a
F Junker et al.
The Biochemical journal, 300 ( Pt 2), 429-436 (1994-06-01)
2-Aminobenzenesulphonic acid (2AS) is degraded by Alcaligenes sp. strain O-1 via a previously detected but unidentified intermediate. A mutant of strain O-1 was found to excrete this intermediate, which was isolated and identified by m.s., 1H- and 13C-n.m.r. as 3-sulphocatechol
Xin-Gui Li et al.
Journal of combinatorial chemistry, 8(2), 174-183 (2006-03-15)
A unique strategy for synthesis of narrowly distributed and inherently self-stabilized copolymer nanoparticles by a simple emulsifier-free polymerization from orthanilic acid and aniline was developed. The polymerization yield, electrical conductivity, size, and its distribution of the nanoparticles could be simultaneously
Determination of nitrite ion and sulfanilic and orthanilic acids by differential pulse polarography.
S T Sulaiman
Analytical chemistry, 56(13), 2405-2407 (1984-11-01)
Inotropic activity of orthanilic and L-cysteic acid on isolated guinea-pig ventricular strips.
F Franconi et al.
Advances in experimental medicine and biology, 217, 159-165 (1987-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service