Skip to Content
Merck
All Photos(1)

Documents

A55500

Sigma-Aldrich

2-Aminofluorene

98%

Synonym(s):

2-Fluorenamine, 2-Fluorenylamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H11N
CAS Number:
Molecular Weight:
181.23
Beilstein:
1945861
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

powder

mp

124-128 °C (lit.)

SMILES string

Nc1ccc-2c(Cc3ccccc-23)c1

InChI

1S/C13H11N/c14-11-5-6-13-10(8-11)7-9-3-1-2-4-12(9)13/h1-6,8H,7,14H2

InChI key

CFRFHWQYWJMEJN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Jun Zhao et al.
Stem cell research & therapy, 10(1), 165-165 (2019-06-15)
Mounting evidence has shown that a novel subset of mesenchymal stem cells (MSCs) derived from human gingiva referred to as gingival mesenchymal stem cells (GMSCs) displays a greater immunotherapeutic potential and regenerative repair expression than MSCs obtained from other tissues.
Olga Rechkoblit et al.
Nature structural & molecular biology, 17(3), 379-388 (2010-02-16)
The aromatic amine carcinogen 2-aminofluorene (AF) forms covalent adducts with DNA, predominantly with guanine at the C8 position. Such lesions are bypassed by Y-family polymerases such as Dpo4 via error-free and error-prone mechanisms. We show that Dpo4 catalyzes elongation from
Ali Bilici et al.
Biomacromolecules, 11(10), 2593-2601 (2010-09-08)
In this paper, the results on horseradish peroxidase (HRP)-catalyzed oxidative polymerization of amine-functionalized fluorene monomer, 2-amino fluorene (AF), are reported. The resulting polymer exhibits an exciting molecular structure and spectral properties. FT-IR and NMR studies show that the two fluorene
Nidhi Jain et al.
Chemical research in toxicology, 21(2), 445-452 (2008-01-16)
Fluorescence spectroscopy was used to study carcinogen-induced conformational heterogeneity in DNA duplexes. The fluorophore 2-aminopurine (AP) was incorporated adjacent (5') to the lesion (G*) in eight different DNA duplexes [d(5'-CTTCT PG* NCCTC-3'):d(5'-GAGGN XTAGAAG-3'), G* = FAF adduct, P = AP
Stephen W Holman et al.
Rapid communications in mass spectrometry : RCM, 22(15), 2355-2365 (2008-07-10)
A 50 m/z unit loss from protonated 4-benzenesulfinyl-3-methylphenylamine has been observed and investigated using electrospray ionisation quadrupole ion trap mass spectrometry (ESI-QIT-MS). It was hypothesised that the specific fragmentation was affected by the presence of an ortho methyl group in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service