Skip to Content
Merck
All Photos(1)

Key Documents

A44509

Sigma-Aldrich

α-Amino-γ-butyrolactone hydrobromide

99%

Synonym(s):

(±)-α-Amino-γ-butyrolactone hydrobromide, DL-Homoserine lactone hydrobromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H7NO2 · HBr
CAS Number:
Molecular Weight:
182.02
Beilstein:
4530084
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

crystalline

color

white to light yellow

mp

220-225 °C (lit.)

application(s)

peptide synthesis

SMILES string

Br[H].NC1CCOC1=O

InChI

1S/C4H7NO2.BrH/c5-3-1-2-7-4(3)6;/h3H,1-2,5H2;1H

InChI key

MKLNTBLOABOJFZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Steven E A Christiaen et al.
Journal of microbiological methods, 87(2), 213-219 (2011-08-23)
A large number of Gram-negative pathogens produce N-acylhomoserine lactones (AHLs) as signal molecules for quorum sensing (QS). This cell-cell communication system allows them to coordinate gene expression and regulate virulence. Therefore, strategies to inhibit QS are promising for the control
Ariel A Romano et al.
Journal of inorganic biochemistry, 107(1), 96-103 (2011-12-20)
Bacteria use small diffusible molecules to exchange information in a process called quorum sensing (QS). An important class of quorum sensing molecules used by Gram-negative bacteria is the family of N-acylhomoserine lactones (HSL). It was recently discovered that a degradation
Cao Xu et al.
Wei sheng wu xue bao = Acta microbiologica Sinica, 51(6), 769-775 (2011-08-27)
Quorum-sensing (QS) is a regulatory mechanism with which bacteria regulate the gene expression according to their population density. Pseudomonas aeruginosa regulates the expression of multiple genes via a hierarchical quorum-sensing cascade through LasR and RhlR and their cognate signal molecules
Fiorela Nievas et al.
Sensors (Basel, Switzerland), 12(3), 2851-2873 (2012-06-28)
Bacteria of the genus Bradyrhizobium are able to establish a symbiotic relationship with peanut (Arachis hypogaea) root cells and to fix atmospheric nitrogen by converting it to nitrogenous compounds. Quorum sensing (QS) is a cell-cell communication mechanism employed by a
Xiaopeng Guo et al.
Wei sheng wu xue bao = Acta microbiologica Sinica, 51(9), 1200-1204 (2011-12-01)
Methanosaeta harundinacea 6Ac displayed a cell-density dependent cell morphology transition between short rods (3 microm - 5 microm) and filaments ( >200 microm), implying a quorum sensing system in this archaeon. This study aimed to confirm that cell morphology related

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service