Skip to Content
Merck
All Photos(2)

Key Documents

A10000

Sigma-Aldrich

Acetone cyanohydrin

99%

Synonym(s):

α-Hydroxyisobutyronitrile, 2-Hydroxyisobutyronitrile

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C(OH)CN
CAS Number:
Molecular Weight:
85.10
Beilstein:
605391
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.02

vapor pressure

0.3 psi ( 20 °C)

Quality Level

Assay

99%

form

liquid

contains

H3PO4 as stabilizer (Europe)
sulfuric acid as stabilizer (US)

expl. lim.

11 %

refractive index

n20/D 1.399 (lit.)

bp

82 °C/23 mmHg (lit.)

mp

−19 °C (lit.)

density

0.932 g/mL at 25 °C (lit.)

SMILES string

CC(C)(O)C#N

InChI

1S/C4H7NO/c1-4(2,6)3-5/h6H,1-2H3

InChI key

MWFMGBPGAXYFAR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Acetone cyanohydrin serves as a source of hydrogen cyanide in the Mitsunobu reaction. Also an intermediate for methacrylamide synthesis.

Application

Acetone cyanohydrin can be used as a cyanide source for:
  • Cyanation reactions of aryl halides, aldehydes and alcohols.
  • Stereospecific epoxide ring opening.
  • Strecker reaction of α-amido sulfones to synthesize α-amino nitriles.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

167.0 °F - closed cup

Flash Point(C)

75 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

H Lauble et al.
Protein science : a publication of the Protein Society, 10(5), 1015-1022 (2001-04-24)
The structure and function of hydroxynitrile lyase from Manihot esculenta (MeHNL) have been analyzed by X-ray crystallography and site-directed mutagenesis. The crystal structure of the MeHNL-S80A mutant enzyme has been refined to an R-factor of 18.0% against diffraction data to
Z I Zholdakova et al.
Gigiena i sanitariia, (8)(8), 16-18 (1994-09-01)
The study of acetone cyanohydrin (ACH) showed that it is an unstable substance. Its stability in water depends on pH of water and less so on its initial concentration; water temperature and biota are insignificant. Acetone and cyanid-ion, forming from
Regiospecific opening of 1, 2-expoxides with acetone cyanohydrin under mildly basic conditions.
Mitchell D and Koenig TM
Tetrahedron Letters, 33(23), 3281-3284 (1992)
Rondon Tosta Ramalho et al.
Acta cirurgica brasileira, 25(1), 111-116 (2010-02-04)
To evaluate the antitumor effect of acetone cyanohydrin in Ehrlich ascites tumor cells in vitro. The Ehrlich ascites tumor cells and lymphocytes were incubated with different concentrations of acetone cyanohydrin (0, 0.5, 1.0, 2.0, 10.0, 20.0 and 30.0 microg x
Cyanohydrins in nature and the laboratory: biology, preparations, and synthetic applications.
Gregory RJH
Chemical Reviews, 99(12), 3649-3682 (1999)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service