935883
TItanium Carbide Mxene
multi-layer powder, 99.9% trace metals basis (exluding Al)
Synonym(s):
Ti3C2 Mxene
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
Quality Level
Assay
99.9% trace metals basis (exluding Al)
form
powder
greener alternative product characteristics
Waste Prevention
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
particle size
10-30 μm
greener alternative category
General description
Two-dimensional (2D) transition-metal carbides, carbonitrides and nitrides, known as MXenes, have attracted significant attention due to their outstanding electronic, ionic, optical, mechanical, and thermal properties, as well as plasmonic behaviour and versatile/tunable surface chemistries [1-4]. We offer TItanium Based MXene multi-layer powder (Ti3C2) with assay purity of 99.9%, and particle size distribuion of 10 ~ 15 μm.
We are committed to bringing you greener alternative products, which adhere to one or more of the 12 Principles of Green Chemistry. This product is synthesized using recycled TiO2 as a source material, and its production process involves reduced processing steps with improved electrical conductivity and enhanced electromagnetic shielding effectiveness. Click here for more information.
Application
Ti3C2 MXenes are suitable for various applications, such as (but not limited to):
- Energy conversion (eg: solar cells, photocatalytic fuel production, such as hydrogen evolution from water splitting, and carbon dioxide reduction) and storage (eg: rechargeable Li-ion and beyond batteries, supercapacitors) systems ,;
- Photocatalytic applications ,;
- Bio-related applications (eg: drug delivery, neuroelectronic devices, photothermal therapy of cancer, biosensors, and tissue engineering),.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
The Rise of MXenes
ACS Nano, 13(8), 8491?8494-8491?8494 (2017)
MXene in the lens of biomedical engineering: synthesis, applications and future outlook
BioMedical Engineering OnLine, 20(1):33 (2021)
Two-Dimensional Ti3C2 MXene for High-Resolution Neural Interfaces
ACS Nano, 12(10), 10419-10429 (2018)
Applications of 2D MXenes in energy conversion and storage systems
Chemical Society Reviews, 48(1), 72-133 (2019)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service