Skip to Content
Merck
All Photos(2)

Key Documents

932019

Sigma-Aldrich

9,9-Bis[4-[(4-ethenylphenyl)methoxy]phenyl]-N2,N7-di-1-naphthalenyl-N2,N7-diphenyl-9H-Fluorene-2,7-diamine

≥98% (HPLC)

Synonym(s):

VB-FNPD

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C75H56N2O2
CAS Number:
Molecular Weight:
1017.26
MDL number:
UNSPSC Code:
12352116
NACRES:
NA.23

Quality Level

Assay

≥98% (HPLC)

solubility

chloroform: soluble
dichloromethane: soluble
toluene: soluble

λmax

354 nm±5 nm in dichloromethane
378 nm±5 nm in dichloromethane

fluorescence

λem 489 nm±10 nm in dichloromethane

Orbital energy

HOMO 5.3 eV 
LUMO 2.3 eV 

Application

9,9-Bis[4-[(4-ethenylphenyl)methoxy]phenyl]-N2,N7-di-1-naphthalenyl-N2,N7-diphenyl-9H-Fluorene-2,7-diamine, also known as VB-FNPD, is a solution-processable crosslinkable Hole Transport / Hole Injection Layer (HTL / HIL) material popularly used in optoelectronic devices, such as quantum-dot light-emitting diodes (QD-LEDs) [1], and perovskite light-emitting diodes (PeLEDs) [2,3].For instance, it was shown that thermally crosslinkable VB-FNPD is a great candidate for the HTL of QD-LEDs [1]. Device with VB-FNPD HTL crosslinking at 170 °C has the highest luminance of 7702 cd/m2, the maximum c.d. (J) of 41.98 mA/cm2, the maximum current efficiency (CE) of 5.45 Cd/A, and the maximum external quantum efficiency (EQE) of 1.64%.VB-FNPD was also successfully used as hole-transporting material in PeLEDs to to improve the hole-injection efficiency of PeLEDs [2,3].

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cross-linked hole transport layers for high-efficiency perovskite tandem solar cells.
Wang Y, et al.
Science China: Chemistry, 64, 2025-2034 (2021)
Chun-Cheng Lin et al.
Polymers, 12(10) (2020-10-03)
In this paper, a thermally crosslinkable 9,9-Bis[4-[(4-ethenylphenyl)methoxy]phenyl]-N2,N7-di-1-naphthalenyl-N2,N7-diphenyl-9H-fluorene-2,7-diamine (VB-FNPD) film served as the hole transporting layer (HTL) of perovskite CsPbBr3 quantum-dot light-emitting diodes (QD-LEDs) was investigated and reported. The VB-FNPD film crosslinked at various temperatures in the range of 100~230 °C
Ming-Chuan Hsiao et al.
Physical chemistry chemical physics : PCCP, 21(15), 7867-7873 (2019-03-28)
Herein, four pyridine-based chelating ligands, including 1,10-phenanthroline, 4,4'-bipyridine, 4,7-diphenyl-1,10-phenanthroline (Bphen) and pyridine, are used as surface treatment agents for improving the electronic properties of perovskite materials. The results of the steady-state photoluminescence (PL), time resolved PL and hole-only devices made

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service