926604
Upconversion Nanoparticles
NaYF4-Yb,Er@NaYF4, PEG-COOH modified core-shell, fluorescence λex 980 nm, green light
Synonym(s):
UCNPs
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352302
NACRES:
NA.23
Recommended Products
Quality Level
concentration
2 mg/mL in water
matrix active group
PEG-COOH surface treatment
particle size
30 nm±5 nm
fluorescence
λex 980 nm (green)
λem 540 nm
λem 650 nm
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
Application
Upconversion nanoparticles (ucnps), are the result of a unique optical process in which near-infrared (NIR) light excitation is converted into visible and/or UV emission. Upconversion (UC) utilizes sequential absorption of multiple photons utilizing both lifetime and real ladder-like energy levels of ions in a host lattice to produce a higher energy anti-Stokes luminescence.
This optical features of UCNPs, results in deep tissue penetration and minimal autofluorescence background, for a broad range of applications of UCNP in diagnostics and biomedical imaging systems.
Applications include:
Fluorescent microscopy
Deep-tissue bioimaging
Nanomedicine
Optogenetics
Security labelling
Volumetric display
This optical features of UCNPs, results in deep tissue penetration and minimal autofluorescence background, for a broad range of applications of UCNP in diagnostics and biomedical imaging systems.
Applications include:
Fluorescent microscopy
Deep-tissue bioimaging
Nanomedicine
Optogenetics
Security labelling
Volumetric display
Legal Information
Product of RuixiBiotechCo.Ltd
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Shihui Wen et al.
Nature communications, 9(1), 2415-2415 (2018-06-22)
Lanthanide-doped upconversion nanoparticles (UCNPs) are capable of converting near-infra-red excitation into visible and ultraviolet emission. Their unique optical properties have advanced a broad range of applications, such as fluorescent microscopy, deep-tissue bioimaging, nanomedicine, optogenetics, security labelling and volumetric display. However
Christoph Drees et al.
Angewandte Chemie (International ed. in English), 55(38), 11668-11672 (2016-08-12)
Upconversion nanoparticles (UCNPs) convert near-infrared into visible light at much lower excitation densities than those used in classic two-photon absorption microscopy. Here, we engineered <50 nm UCNPs for application as efficient lanthanide resonance energy transfer (LRET) donors inside living cells. By
Upconversion nanoparticles: design, nanochemistry, and applications in theranostics.
Guanying Chen et al.
Chemical reviews, 114(10), 5161-5214 (2014-03-13)
Yujia Liu et al.
Nature, 543(7644), 229-233 (2017-02-23)
Lanthanide-doped glasses and crystals are attractive for laser applications because the metastable energy levels of the trivalent lanthanide ions facilitate the establishment of population inversion and amplified stimulated emission at relatively low pump power. At the nanometre scale, lanthanide-doped upconversion
Daniel J Gargas et al.
Nature nanotechnology, 9(4), 300-305 (2014-03-19)
Imaging at the single-molecule level reveals heterogeneities that are lost in ensemble imaging experiments, but an ongoing challenge is the development of luminescent probes with the photostability, brightness and continuous emission necessary for single-molecule microscopy. Lanthanide-doped upconverting nanoparticles overcome problems
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service