Skip to Content
Merck
All Photos(1)

Key Documents

923087

Sigma-Aldrich

Copper(II) nitrate hydrate

≥99.9% trace metals basis

Synonym(s):

Cupric nitrate hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cu(NO3)2 · xH2O
CAS Number:
Molecular Weight:
187.56 (anhydrous basis)
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.23
grade:
for analytical purposes
form:
crystals and lumps
Pricing and availability is not currently available.

grade

for analytical purposes

Quality Level

Assay

≥99.9% trace metals basis

form

crystals and lumps

mp

114.5 °C (dec.) (lit.)

storage temp.

2-8°C

SMILES string

O.[Cu++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/Cu.2NO3.H2O/c;2*2-1(3)4;/h;;;1H2/q+2;2*-1;

InChI key

VMKYLARTXWTBPI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Cu(NO3)2 · xH2O is a promising anode material for lithium ion batteries. Some studies indicate high initial discharge capacity of upto 2200 mAhg-1.[1][2][3]It is also used as an oxidizing agent for the one step synthesis of CuO/Cu2O/C anode for Li-ion batteries, another promising anode material for Li-ion batteries.[4]

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Ox. Sol. 2 - Skin Corr. 1B

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Copper nitrate hydrate as novel high capacity anode material for lithium-ion batteries.
Wu K, et al.
Journal of Power Sources, 248, 205-211 (2014)
Copper-Based Nanomaterials for High-Performance Lithium-Ion Batteries.
Xu J, et al.
Particle & Particle Systems Characterization, 33(11), 784-810 (2016)
One-step solution combustion synthesis of CuO/Cu2O/C anode for long cycle life Li-ion batteries.
Xu C, et al.
Carbon, 142, 51-59 (2019)
Lithium storage mechanism in superior high capacity copper nitrate hydrate anode material.
Jiang X, et al.
Journal of Power Sources, 260, 218-224 (2014)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service