Skip to Content
Merck
All Photos(1)

Key Documents

920398

Sigma-Aldrich

Lithium bis(trimethylsilyl)amide

99.9% trace metals basis

Synonym(s):

LHMDS, LiHMDS, LiTMSA, Lithium hexamethyldisilazide, Hexamethyldisilazane lithium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)3Si]2NLi
CAS Number:
Molecular Weight:
167.33
MDL number:
UNSPSC Code:
12352111
form:
solid

Quality Level

Assay

99.9% trace metals basis

form

solid

density

0.860 g/mL at 25 °C (lit.)

application(s)

battery manufacturing

SMILES string

[Li]N([Si](C)(C)C)[Si](C)(C)C

InChI

1S/C6H18NSi2.Li/c1-8(2,3)7-9(4,5)6;/h1-6H3;/q-1;+1

InChI key

YNESATAKKCNGOF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Lithium bis(trimethylsilyl)amide also known as lithium hexamethyldisilazide (LiHMDS) is a non-nucleophilic strong base. It exhibits ionic conductivity and is widely used as a lithium source and additive in electrolyte formulations for lithium-ion batteries.

Application

Lithium bis(trimethylsilyl)amide can be used:


  • As an electrolyte additive for non-aqueous lithium metal batteries. LiHMDS acts as a scavenger for hydrofluoric acid and forms an electrochemical robust cathode|electrolyte interphase (CEI) and suppresses the side reactions with the electrolyte solution.
  • As a lithium precursor for atomic layer deposition(ALD) of textured Li4Ti5O12 as anode material for Li-ion ultrafast charging thin-film batteries. It enables the controlled delivery of lithium atoms into the deposition process, leading to the growth of thin films with precise thickness and composition.
  • As a precursor to fabricate in situ lithiated quinone cathode as high-capacity organic electrode material for all-solid-state thin-film battery setup.

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Sol. 1 - Self-heat. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 2

Flash Point(F)

62.6 °F - closed cup

Flash Point(C)

17 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lithium Phosphate Thin Films Grown by Atomic Layer Deposition.
Hamalainen J, et al.
Journal of the Electrochemical Society, 159(3), A259-A259 (2012)
Lithium hexamethyldisilazide as electrolyte additive for efficient cycling of high-voltage non-aqueous lithium metal batteries
Danfeng Zhang, et al.
Nature Communications, 13, 6966-6966 (2022)
Atomic layer deposition of textured Li4Ti5O12: a high-power and long-cycle life anode for lithium-ion thin-film batteries
Jan Speulmanns, et al
Small, 17, 2102635-2102635 (2021)
On the Stability of LiFePO4 Olivine Cathodes under Various Conditions (Electrolyte Solutions, Temperatures)
Koltypin M, et al.
Electrochemical and Solid-State Letters, 10(2) (2007)
Maksym Yarema et al.
ACS nano, 5(5), 3758-3765 (2011-04-20)
Here, we present a hot injection synthesis of colloidal Ag chalcogenide nanocrystals (Ag(2)Se, Ag(2)Te, and Ag(2)S) that resulted in exceptionally small nanocrystal sizes in the range between 2 and 4 nm. Ag chalcogenide nanocrystals exhibit band gap energies within the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service