Skip to Content
Merck
All Photos(1)

Key Documents

915939

Sigma-Aldrich

Lithium titanate

spinel "LTO" powder, battery grade

Synonym(s):

LTO nanopowder, Lithium titanium oxide, NANOMYTE® BE-10, LTO, Lithium titanate spinel oxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Li4Ti5O12
MDL number:
UNSPSC Code:
12352303
NACRES:
NA.23

grade

battery grade

Quality Level

form

powder

mol wt

Mw 459.09 g/mol

composition

Li4Ti5O12

particle size

1-3 μm

density

3.48 g/cm3 (lit.)

application(s)

battery manufacturing

SMILES string

[Li+].[Li+].[Li+].[Li+].[O-][Ti](=O)O[Ti](O[Ti]([O-])=O)(O[Ti]([O-])=O)O[Ti]([O-])=O

InChI

1S/4Li.12O.5Ti/q4*+1;;;;;;;;;4*-1;;;;;

InChI key

BNQVSKURWGZJMY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Lithium-titanate battery is a kind of new lithium-ion batteries, and it can be charged by high current, but changes in temperature and capacity have a great influence on the battery performance. The battery stability and the charging curve are examined in this paper for the high current and various test conditions. It is found that the LTO has an advanced performance in temperature rise.

Legal Information

NANOMYTE is a registered trademark of NEI Corporation

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

John B Goodenough et al.
Journal of the American Chemical Society, 135(4), 1167-1176 (2013-01-09)
Each cell of a battery stores electrical energy as chemical energy in two electrodes, a reductant (anode) and an oxidant (cathode), separated by an electrolyte that transfers the ionic component of the chemical reaction inside the cell and forces the
Lithium Titanate-Based Lithium-Ion Batteries.
Liu J, et al.
Wiley-VCH (2020)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service