Skip to Content
Merck
All Photos(1)

Key Documents

915114

Sigma-Aldrich

Lithium tin phosphorus sulfide (LSPS)

battery grade

Synonym(s):

Nanomyte SSE-10

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Li10SnP2S12
Molecular Weight:
634.85
UNSPSC Code:
12352300
NACRES:
NA.23

grade

battery grade

Quality Level

Assay

≥99.5%

form

powder

mol wt

Mw 634.85 g/mol

composition

Li10SnP2S12

surface area

1.45 m2/g

particle size

7-8 μm (average)

conductivity

0.1-10  mS/cm (ionic conductivity, cold-pressed pellet at room temperature)

density

2.254 g/cm3 (lit.)

application(s)

battery manufacturing

Application

Solid electrolytes (SEs) have become a practical option for lithium ion and lithium metal batteries because of their improved safety over commercially available ionic liquids. Solid electrolyte materials are highly conductive and made for advanced solid-state lithium ion battery technologies. These solutions are high purity and battery grade thus making them suitable as standards in LIB research

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Aquatic Acute 1 - Eye Dam. 1 - Flam. Sol. 1 - Skin Corr. 1B - Water-react 1

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service