Skip to Content
Merck
All Photos(1)

Key Documents

912034

Sigma-Aldrich

Trimethyl chitosan

high molecular weight, degree of quaternization 30-70%

Synonym(s):

N,N,N-Trimethyl chitosan, Chitosan trimethyl, Mucoadhesive polymers, TMC Chitosan

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C9H18ClNO4)n
CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

Quality Level

form

powder

color

light yellow to light brown

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Chitosan offers remarkable biological properties, it has been widely used in drug delivery and tissue engineering applications. Chitosan has good mucoadhesive properties due to its positive charge, which increases the adhesion to mucosa and facilitates drug penetration. Also, chitosan possesses hemostatic properties, which makes it a good candidate for wound dressing applications.

The quaternization of the primary amine increases the water solubility of chitosan and keeps chitosan soluble over a wide pH range. Among all the quaternized chitosans, N, N, N-trimethyl chitosan chloride (TMC) is the most widely applied in biomedical applications.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation and Characterization of Particles from Chitosan with Different Molecular Weights and Their Trimethyl Chitosan Derivatives for Nasal Immunization.
Boonyo W, et al.
Journal of Metals, Materials and Minerals, 18(2), 59-65 (2008)
Mohamed H Ramadan et al.
Langmuir : the ACS journal of surfaces and colloids, 30(25), 7485-7495 (2014-06-04)
We report a thermoresponsive chemical modification strategy of hyaluronic acid (HA) for coating onto a broad range of biomaterials without relying on chemical functionalization of the surface. Poly(di(ethylene glycol) methyl ether methacrylate) (PMEO2MA), a polymer with a lower critical solution
Rajesh Kumar Kainthan et al.
Biomacromolecules, 9(3), 886-895 (2008-02-06)
This paper discusses the binding and release properties of hydrophobically modified hyperbranched polyglycerol-polyethylene glycol copolymers that were originally developed as human serum albumin (HSA) substitutes. Their unimolecular micellar nature in aqueous solution has been proven by size measurements and other
Jameel Ahmad Khan et al.
Biomacromolecules, 7(5), 1386-1388 (2006-05-09)
Recent advances in understanding biological systems have proven that RNA is not merely the carrier of genetic information, but also a key molecule in regulation of gene expression and other crucial metabolic processes. Therefore, it is being considered as an
Preparation and NMR characterization of highly substituted IV-trimethyl chitosan chloride.
Sieval A B, et al.
Carbohydrate Polymers, 36, 157-165 (1998)

Articles

Professor Robert K. Prud’homme introduces flash nanoprecipitation (FNP) for nanoparticle fabrication, which is a scalable, rapid mixing process for nanoparticle formulations.

Professor Robert K. Prud’homme introduces flash nanoprecipitation (FNP) for nanoparticle fabrication, which is a scalable, rapid mixing process for nanoparticle formulations.

Professor Robert K. Prud’homme introduces flash nanoprecipitation (FNP) for nanoparticle fabrication, which is a scalable, rapid mixing process for nanoparticle formulations.

Professor Robert K. Prud’homme introduces flash nanoprecipitation (FNP) for nanoparticle fabrication, which is a scalable, rapid mixing process for nanoparticle formulations.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service