Skip to Content
Merck
All Photos(2)

Key Documents

907294

Sigma-Aldrich

Fmoc-L-Photo-Phe-OH

≥95%

Synonym(s):

(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(4-(3-(trifluoromethyl)-3H-diazirin-3-yl)phenyl)propanoic acid, N-α-(9-Fluorenylmethyloxycarbonyl)-4-(trifluoromethyldiazirin)-L-phenylalanine, Diazirine amino acid, Fmoc-Tdf-OH, Photo-Phe, Photo-crosslinking amino acid, Photoprobe building block

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H20F3N3O4
CAS Number:
Molecular Weight:
495.45
MDL number:
UNSPSC Code:
12352209
NACRES:
NA.22

Assay

≥95%

form

powder

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

−20°C

SMILES string

FC(F)(F)C5(N=N5)c1ccc(cc1)C[C@H](NC(=O)OCC2c3c(cccc3)c4c2cccc4)C(=O)O

InChI

1S/C26H20F3N3O4/c27-26(28,29)25(31-32-25)16-11-9-15(10-12-16)13-22(23(33)34)30-24(35)36-14-21-19-7-3-1-5-17(19)18-6-2-4-8-20(18)21/h1-12,21-22H,13-14H2,(H,30,35)(H,33,34)/t22-/m0/s1

InChI key

UHXIAQUVGJCYSA-QFIPXVFZSA-N

Application

Fmoc-L-Photo-Phe-OH is a diazirine-containing, Fmoc-protected phenylalanine amino acid and multifunctional photo-crosslinker. Its incorporation into peptides or small-molecule probes and tools allows for photoaffinity labeling of cellular targets and protein-protein interactions upon UV light (∼360 nm) irradiation to form a covalent bond. This and other multifunctional probe building blocks will continue to accelerate drug discovery research for probing cellular mechanisms, target ID/validation, and understanding traditionally undruggable targets. An unprotected version is also available as 907340.

Product can be used with our line of photoreactors: Including Penn PhD (Z744035) & SynLED 2.0 (Z744080)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ryuto Kino et al.
Bioorganic & medicinal chemistry letters, 25(15), 2972-2975 (2015-06-06)
Inhibition of amyloid-β (Aβ) aggregation could be a drug development target for treating Alzheimer disease. Insufficient activity to inhibit aggregation, however, remains a key issue. Here, we report a covalent modifier-type aggregation inhibitor of Aβ, diazirine-equipped cyclo-KLVF(β-Ph)F (2). Due to
Michiel A Leeuwenburgh et al.
Organic letters, 8(8), 1705-1708 (2006-04-07)
[reaction: see text] A novel solid-phase synthesis strategy toward succinylhydroxamate peptides, using an appropriately protected hydroxamate building block, is described. Rapid and efficient access is gained to amine-functionalized peptides, which can be decorated with, for instance, a fluorescent label. In
Dany Fillion et al.
Journal of medicinal chemistry, 49(7), 2200-2209 (2006-03-31)
A stereospecific convergent synthesis of N-[(9-fluorenyl)methoxycarbonyl]-p-[3-(trifluoromethyl)-3H-diazirin-3-yl]-l-phenylalanine (Fmoc-12, Fmoc-Tdf) and its incorporation into the C-terminal position of the angiotensin II (AngII) peptide to form (125)I[Sar(1),Tdf(8)]AngII ((125)I-13) is presented. This amino acid photoprobe is a highly reactive carbene-generating diazirine phenylalanine derivative that
R Falchetto et al.
The Journal of biological chemistry, 266(5), 2930-2936 (1991-02-15)
A synthetic, 28-residue peptide derived from the calmodulin-binding sequence of the plasma membrane Ca2+ pump (C28W) inhibits the ATPase activity of a calpain-produced, truncated fragment of the enzyme. The fragment, which has lost the calmodulin-binding domain, has a molecular mass
M Ploug et al.
Biochemistry, 37(11), 3612-3622 (1998-04-02)
Binding of urokinase-type plasminogen activator (uPA) to its cellular receptor (uPAR) renders the cell surface a favored site for plasminogen activation. Recently, a 15-mer peptide antagonist of the uPA-uPAR interaction, with an IC50 value of 10 nM, was identified using

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service