Skip to Content
Merck
All Photos(1)

Key Documents

906379

Sigma-Aldrich

COi8DFIC

≥98%

Synonym(s):

2,2′-[[4,4,11,11-tetrakis(4-hexylphenyl)-4,11-dihydrothieno[2′,3′:4,5]thieno[2,3-d]thieno[2′′′′,3′′′′:4′′′,5′′′]thieno[2′′′,3′′′:4′′,5′′]pyrano[2′′,3′′:4′,5′]thieno[2′,3′:4,5]thieno[3,2-b]pyran-2,9-diyl]bis[methylidyne(5,6-difluoro, NFA146, O6T-4F, PCE146

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C94H76F4N4O4S6
CAS Number:
Molecular Weight:
1594.02
UNSPSC Code:
12352101
NACRES:
NA.23

description

Band gap: 1.62 eV

Assay

≥98%

form

solid

solubility

soluble (chloroform, CB and ODCB)

Orbital energy

HOMO -5.5 eV 
LUMO -3.88 eV 

General description

COi8DFIC or O6T-4F is a highly efficient, n-type, low-bandgap nonfullerene acceptor with strong NIR absorption.
In a recent study, COi8DFIC or O6T-4F was selected in a Tandem cell by computer assited design and gave a record PCE of 17.3∃% for fabricated organic solar cells.
COi8DFIC or O6T-4F is frequently selected to blend with a narrow-bandgap donor material and another narrow bandgap acceptor material to fabricate ternary organic solar cells. The PTB7-Th:COi8DFIC:PC71BM ternary cells offered a PCE of 14.08%. By further adopting a post-annealing process, an outstanding PCE of 14.62% can be achieved. Furthermore, the device utilizing COi8DFIC exhibited a good thermal stability with PCEs over 13.5% in a wide temperature range (70–160 °C).

Application

COi8DFIC is primarily utilized as a non-fullerene acceptor in OPV devices. It exhibits a broad absorption spectrum, enabling it to absorb light across a wide range of wavelengths, including the visible and near-infrared regions. This property allows for efficient utilization of a broader range of solar radiation, enhancing the light-harvesting capability of the OPV device. COi8DFIC can be employed as the electron transport material in OFET devices.
COi8DFIC or O6T-4F is a highly efficient, n-type, low-bandgap nonfullerene acceptor with strong NIR absorption
In a recent study, COi8DFIC or O6T-4F was selected in a Tandem cell by computer assited design and gave a record PCE of 17.3% for fabricated organic solar cells.

Tandem Cell Device performance:
ITO/ZnO/PFN-Br/PBDB-T:F-M/M-PEDOT/ZnO/PTB7- Th:O6T-4F:PC71BM/MoO3/Ag
Voc=1.642 V
Jsc=14.35 mA/cm2
FF=73.7%
PCE=17.3%
COi8DFIC or O6T-4F is frequently selected to blend with a narrow-bandgap donor material and another narrow bandgap acceptor material to fabricate ternary organic solar cells. The PTB7-Th:COi8DFIC:PC71BM ternary cells offered a PCE of 14.08%. By further adopting a post-annealing process, an outstanding PCE of 14.62% can be achieved. Furthermore, the device utilizing COi8DFIC exhibited a good thermal stability with PCEs over 13.5% in a wide temperature range (70-160 °C).

Device structure:
ITO/ZnO/PTB7-Th:COi8DFIC:PC71BM/MoO3/Ag
  • Before annealing
Voc=0.702 V
Jsc=27.74 mA/cm2
FF=0.701
PCE=13.65%

  • After annealing at 80°C
Voc=0.727 V
Jsc=27.39 mA/cm2
FF=0.734
PCE=14.62%
COi8DFIC, an efficient non-fullerene acceptor material, has a strong near-infrared range (NIR) light absorption. It can be used as an n-type small molecule acceptor material for the fabrication of polymeric solar cells.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ternary organic solar cells offer 14% power conversion efficiency.
Xiao Z, et al.
Science Bulletin, 62(23), 1562-1564 (2017)
Simultaneously improved efficiency and average visible transmittance of semitransparent polymer solar cells with two ultra-narrow bandgap nonfullerene acceptors
Ma X, et al.
Journal of Material Chemistry A, 6(43), 21485-21492 (2018)
Thermostable single-junction organic solar cells with a power conversion efficiency of 14.62 %.
Li H, et al.
Science Bulletin, 63(6), 340-342 (2018)
Molecular order control of non-fullerene acceptors for high-efficiency polymer solar cells
Li W, et al.
Joule, 3(3), 819-833 (2019)
Comparative analysis of burn-in photo-degradation in non-fullerene COi8DFIC acceptor based high-efficiency ternary organic solar cells
Duan L, et al.
Materials Chemistry Frontiers., 3(6), 1085-1096 (2019)

Articles

Professor Chen (Nankai University, China) and his team explain the strategies behind their recent record-breaking organic solar cells, reaching a power conversion efficiency of 17.3%.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service