All Photos(3)
About This Item
Empirical Formula (Hill Notation):
C40H34F12IrN4P
CAS Number:
Molecular Weight:
1021.90
MDL number:
UNSPSC Code:
12161600
NACRES:
NA.22
Recommended Products
form
solid
reaction suitability
core: iridium
reagent type: catalyst
reaction type: Photocatalysis
mp
>300 °C
photocatalyst activation
465 nm
SMILES string
FC1=C(C2=NC=C(F)C=C2)C([Ir]3([N]4=CC=C(C(C)(C)C)C=C4C5=CC(C(C)(C)C)=CC=[N]53)C6=CC(F)=CC(F)=C6C7=CC=C(F)C=N7)=CC(F)=C1.FP(F)(F)(F)(F)F
Application
Ir(dFFppy)2(dtbbpy)PF6 is a versatile photocatalyst that can be used for a variety of synthetic transformations.
Product can be used with our line of photoreactors: Including Penn PhD (Z744035) & SynLED 2.0 (Z744080)
Product can be used with our line of photoreactors: Including Penn PhD (Z744035) & SynLED 2.0 (Z744080)
related product
Product No.
Description
Pricing
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Craig P Johnston et al.
Nature, 536(7616), 322-325 (2016-08-19)
In the past 50 years, cross-coupling reactions mediated by transition metals have changed the way in which complex organic molecules are synthesized. The predictable and chemoselective nature of these transformations has led to their widespread adoption across many areas of
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service