Skip to Content
Merck
All Photos(1)

Key Documents

901074

Sigma-Aldrich

Ir(MDQ)2(acac)

sublimed

Synonym(s):

Bis(2-methyldibenzo[f,h]quinoxaline)(acetylacetonate)iridium(III)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C39H29N4O2Ir
CAS Number:
Molecular Weight:
777.89
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

Quality Level

Assay

≥99% (sublimed, HPLC)

form

powder

color

red

SMILES string

CC1=NC(C(C=CC=C2)=C2C3=C4C5=CC=C3)=C4[N]([Ir]56OC(C)C=C(C)O6)=C1

InChI

1S/C17H11N2.C5H9O2.Ir/c1-11-10-18-16-14-8-4-2-6-12(14)13-7-3-5-9-15(13)17(16)19-11;1-4(6)3-5(2)7;/h2-7,9-10H,1H3;3-4,7H,1-2H3;/q;-1;+2/p-1/b;5-3-;

InChI key

FQHZITKUOOFGDL-FMFSYIAASA-M

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Matthew J Jurow et al.
Nature materials, 15(1), 85-91 (2015-10-06)
Controlling the alignment of the emitting molecules used as dopants in organic light-emitting diodes is an effective strategy to improve the outcoupling efficiency of these devices. To explore the mechanism behind the orientation of dopants in films of organic host
Sebastian Reineke et al.
Nature, 459(7244), 234-238 (2009-05-16)
The development of white organic light-emitting diodes (OLEDs) holds great promise for the production of highly efficient large-area light sources. High internal quantum efficiencies for the conversion of electrical energy to light have been realized. Nevertheless, the overall device power
Novel distorted donor-acceptor type deep blue fluorescent emitter for high efficiency in non-doped blue and cool white organic light-emitting diodes.
Choi JM, et al.
Dyes and Pigments, 142, 243-248 (2017)

Articles

Professor Shinar highlights low-cost, disposable sensor configurations in organic and hybrid electronics for healthcare applications.

Professor Shinar highlights low-cost, disposable sensor configurations in organic and hybrid electronics for healthcare applications.

Professor Shinar highlights low-cost, disposable sensor configurations in organic and hybrid electronics for healthcare applications.

Professor Shinar highlights low-cost, disposable sensor configurations in organic and hybrid electronics for healthcare applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service