Skip to Content
Merck
All Photos(1)

Documents

89761

Sigma-Aldrich

N-Boc- 2,2′-(ethylenedioxy)diethylamine

≥95.0% (NT), for peptide synthesis

Synonym(s):

N-Boc-3,6-dioxa-1,8-octanediamine, tert-Butyl 2-[2-(2-aminoethoxy)ethoxy]ethylcarbamate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H24N2O4
CAS Number:
Molecular Weight:
248.32
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

product name

N-Boc- 2,2′-(ethylenedioxy)diethylamine, ≥95.0% (NT)

Quality Level

Assay

≥95.0% (NT)

form

liquid

reaction suitability

reagent type: cross-linking reagent

density

1.046 g/mL at 20 °C

functional group

Boc
amine

SMILES string

NCCOCCOCCNC(OC(C)(C)C)=O

InChI

1S/C11H24N2O4/c1-11(2,3)17-10(14)13-5-7-16-9-8-15-6-4-12/h4-9,12H2,1-3H3,(H,13,14)

InChI key

OCUICOFGFQENAS-UHFFFAOYSA-N

Application

Linker reagent for the prep. of a broad variety of linked tags, monomers for polymerisation, ionophores etc., e.g. biotinylation reagents.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Braun, M., et al.
European Journal of Organic Chemistry, 7, 1173 - 1182 (2000)
Sigal, G. B., et al.
Journal of the American Chemical Society, 118, 3789-3800 (1996)
Michelle Trester-Zedlitz et al.
Journal of the American Chemical Society, 125(9), 2416-2425 (2003-02-27)
A method is described for the elucidation of protein-protein interactions using novel cross-linking reagents and mass spectrometry. The method incorporates (1) a modular solid-phase synthetic strategy for generating the cross-linking reagents, (2) enrichment and digestion of cross-linked proteins using microconcentrators
Spinke, J., et al.
The Journal of Physical Chemistry, 99(9), 7012-7019 (1993)

Articles

Polyethylene glycol (PEG) reagents offer favorable characteristics for various applications, including biocompatibility and solubility.

Polyethylene glycol (PEG) reagents offer favorable characteristics for various applications, including biocompatibility and solubility.

Polyethylene glycol (PEG) reagents offer favorable characteristics for various applications, including biocompatibility and solubility.

Polyethylene glycol (PEG) reagents offer favorable characteristics for various applications, including biocompatibility and solubility.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service