Skip to Content
Merck
All Photos(3)

Key Documents

861677

Sigma-Aldrich

DL-Cysteine

for peptide synthesis

Synonym(s):

(±)-2-Amino-3-mercaptopropionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HSCH2CH(NH2)COOH
CAS Number:
Molecular Weight:
121.16
Beilstein:
1721406
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

Product Name

DL-Cysteine, technical grade

grade

technical grade

Quality Level

form

powder

reaction suitability

reaction type: solution phase peptide synthesis

mp

225 °C (dec.) (lit.)

application(s)

peptide synthesis

SMILES string

NC(CS)C(O)=O

InChI

1S/C3H7NO2S/c4-2(1-7)3(5)6/h2,7H,1,4H2,(H,5,6)

InChI key

XUJNEKJLAYXESH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

NMDA agonist that is an agonist at quisqualate receptors at high concentration.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kun Ye et al.
Journal of materials chemistry. B, 7(31), 4794-4800 (2019-08-08)
Alkaline phosphatase (ALP) is extensively used as a clinical biomarker because of its close relevance with a variety of diseases. Thus, exploring reliable and practical methods for ALP analysis is of great significance. In the present work, we explored MIL-53(Fe)
Armand O Brown et al.
mBio, 10(4) (2019-07-04)
Enterococcus faecalis, a Gram-positive bacterium, and Candida albicans, a polymorphic fungus, are common constituents of the microbiome as well as increasingly problematic causes of infections. Interestingly, we previously showed that these two species antagonize each other's virulence and that E.
Christoph Bock et al.
Scientific reports, 9(1), 11884-11884 (2019-08-17)
The lysosomal polypeptide transporter TAPL belongs to the superfamily of ATP-binding cassette transporters. TAPL forms a homodimeric transport complex, which translocates oligo- and polypeptides into the lumen of lysosomes driven by ATP hydrolysis. Although the structure and the function of
Gongke Wang et al.
Materials science & engineering. C, Materials for biological applications, 103, 109856-109856 (2019-07-28)
Protein coronas provide a novel technique for the bio identification of nanoparticles in physiological environments, to further elucidate the biological effects of nanoparticles in biomedical applications. Herein, we investigated the adsorption of bovine serum albumin (BSA) on gold nanoparticles (AuNPs)
Jianbo Yang et al.
Melanoma research, 29(4), 365-375 (2019-05-30)
Chondroitin sulfate proteoglycan 4 (CSPG4) is a cell surface proteoglycan that enhances malignant potential in melanoma and several other tumor types. CSPG4 functions as a transmembrane scaffold in melanoma cells to activate oncogenic signaling pathways such as focal adhesion kinase

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service