Skip to Content
Merck

809691

Sodium taurochenodeoxycholate-2,2,3,4,4,6,6,7,8-d9

≥98 atom % D, ≥98% (CP)

Synonym(s):

Taurochenodeoxycholic-2,2,3,4,4,6,6,7,8-d9 acid sodium salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C26D9H35NaNO6S
Molecular Weight:
530.74
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.12
Isotopic purity:
≥98 atom % D
Assay:
≥98% (CP)
Mass shift:
M+9
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


isotopic purity

≥98 atom % D

Quality Segment

assay

≥98% (CP)

form

powder

technique(s)

mass spectrometry (MS): suitable

mass shift

M+9

SMILES string

O=C(NCCS(=O)(O[Na])=O)CC[C@@H](C)[C@@H]1CCC2C3([2H])[C@@](O)([2H])C([2H])([2H])C4C([2H])([2H])[C@@](O)([2H])C([2H])([2H])C[C@]4(C)C3CC[C@@]21C

InChI

1S/C26H45NO6S.Na/c1-16(4-7-23(30)27-12-13-34(31,32)33)19-5-6-20-24-21(9-11-26(19,20)3)25(2)10-8-18(28)14-17(25)15-22(24)29;/h16-22,24,28-29H,4-15H2,1-3H3,(H,27,30)(H,31,32,33);/q;+1/p-1/t16-,17?,18-,19?,20?,21?,22-,24?,25+,26-;/m1./s1/i8D2,14D2,15D2,18D,22D,24D;

InChI key

IYPNVUSIMGAJFC-YHOAHADESA-M



Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library