808709
Gold nanoparticles
10 nm diameter, biotin terminated, PEG 5000, OD 50, dispersion in H2O
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
PEG 5000
Quality Level
form
dispersion in H2O
nanoparticles
composition
, 2.99E+14 particles/mL
packaging
pkg of 0.5 mL
concentration
3.04 mg/mL in H2O
OD
50
surface area
30.8 m2/g
diameter
10 nm
pH
5-7
density
1.00 g/cm3
λmax
515-520 nm
functional group
biotin
storage temp.
2-8°C
InChI
1S/Au
InChI key
PCHJSUWPFVWCPO-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Gold nanoparticles PEG 5000 biotin terminated.
Application
Biotin functionalized gold nanoparticles are suitable for binding of streptavidin labelled conjugates. The gold nanoparticle label allows for straightforward detection of streptavidin by measuring the increase in absorbance of the sample upon binding. The absorption maximum is gold nanoparticle size dependent, and ranges from 515-520 nm for a 5 nm particle to 572 nm for a 100 nm particle.
Gold nanoparticles 10 nm diameter, biotin terminated, PEG 5000, OD 50, dispersion in H2O can be used as a theranostic agent for potential usage in cancer therapy and treatment of COVID-19. It can also be used in the detection of pathogens for biosensing applications. AuNPs may be used to prepare SARS-CoV-2 conjugate for COVID-19 detection using lateral flow immunoassay technique.
Legal Information
Product of Cytodiagnostics, Inc.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Gold nanoparticles surface-functionalized with paclitaxel drug and biotin receptor as theranostic agents for cancer therapy
Biomaterials, 33(3), 856-866 (2012)
Development and clinical application of a rapid IgM-IgG combined antibody test for SARS-CoV-2 infection diagnosis
Journal of Medical Virology, 33(6), 666-680 (2020)
Colorimetric biosensing of pathogenss using gold nanoparticles
Biotechnology Advances, 33(6), 666-680 (2015)
Optimizing use of theranostic nanoparticles as a life-saving strategy for treating COVID-19 patients
Theranostics, 10(13), 5932-5932 (2020)
Localized surface plasmon resonance (LSPR) of polyelectrolyte-functionalized gold-nanoparticles for bio-sensing
Colloids and Surfaces. A, Physicochemical and Engineering Aspects, 332(2-3), 172-179 (2009)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service