Skip to Content
Merck
All Photos(2)

Key Documents

805203

Sigma-Aldrich

FK 102 Co(III) TFSI salt

Synonym(s):

tris(2-(1H-pyrazol-1-yl)pyridine)cobalt(III) tri[bis(trifluoromethane)sulfonimide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H21CoN12O12S6F18
Molecular Weight:
1334.86
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

98%

Quality Level

form

powder

mp

194-199 °C

SMILES string

O=S([N-]S(=O)(C(F)(F)F)=O)(C(F)(F)F)=O.O=S([N-]S(=O)(C(F)(F)F)=O)(C(F)(F)F)=O.O=S([N-]S(=O)(C(F)(F)F)=O)(C(F)(F)F)=O.N1(C2=NC=CC=C2)N=CC=C1.C3(N4C=CC=N4)=CC=CC=N3.C5(N6C=CC=N6)=CC=CC=N5.[Co+3]

InChI

1S/3C8H7N3.3C2F6NO4S2.Co/c3*1-2-5-9-8(4-1)11-7-3-6-10-11;3*3-1(4,5)14(10,11)9-15(12,13)2(6,7)8;/h3*1-7H;;;;/q;;;3*-1;+3

InChI key

ILXRZLQXWLMDFQ-UHFFFAOYSA-N

General description

FK 102 Co(III) TFSI salt is a cobalt(III) complex that can be used as a p-type dopant to control the type and density of charge carriers in both organic and inorganic semiconductors. The solubility of TFSI allows an increase in the doping potential for hole-conductors in electrochemical devices.

Application

FK 102 Co(III) TFSI salt can be majorly used in the fabrication of dye sensitized solar cells (DSSCs) and perovskite based solar cells (PSCs).
Use this cobalt complexes to increase photovoltages of liquid electrolyte cells substantially or to achieve ultrahigh performance with solid state photovoltaic devices. FK102 cobalt complexes offer guaranteed performance, high reproducibility, consistent results, and are of highest purity. In comparison to triiodide-based redox electrolytes, cobalt complexes in general increase photovoltages and particularly at lower light levels(e.g. for indoor applications), significantly increase device power output.
Recommended use:
In liquid-based electrolytes: typically 0.15-0.2 M of Co(II) and ca. 0.05 M Co(II)
In solid-state photovoltaic cells: up to 10 weight % added to the hole transport material system.

Legal Information

Product of Greatcell Solar Materials Pty Ltd. Greatcell Solar is a registered trademark of Greatcell Solar Materials Pty Ltd.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cooperative tin oxide fullerene electron selective layers for high-performance planar perovskite solar cells
Ke W, et al.
Journal of Material Chemistry A, 4(37), 14276-14283 (2016)
Planar heterojunction perovskite solar cell based on CdS electron transport layer
Abulikemu M, et al.
Thin Solid Films, 636(37), 512-518 (2017)
Co (III) complexes as p-dopants in solid-state dye-sensitized solar cells
Burschka J, et al.
Chemistry of Materials, 25(15), 2986-2990 (2013)
B P MacLeod et al.
Science advances, 6(20), eaaz8867-eaaz8867 (2020-05-20)
Discovering and optimizing commercially viable materials for clean energy applications typically takes more than a decade. Self-driving laboratories that iteratively design, execute, and learn from materials science experiments in a fully autonomous loop present an opportunity to accelerate this research
Julian Burschka et al.
Nature, 499(7458), 316-319 (2013-07-12)
Following pioneering work, solution-processable organic-inorganic hybrid perovskites-such as CH3NH3PbX3 (X = Cl, Br, I)-have attracted attention as light-harvesting materials for mesoscopic solar cells. So far, the perovskite pigment has been deposited in a single step onto mesoporous metal oxide films

Articles

Next generation solar cells have the potential to achieve conversion efficiencies beyond the Shockley-Queisser (S-Q) limit while also significantly lowering production costs.

Dr. Perini and Professor Correa-Baena discuss the latest research and effort to obtain higher performance and stability of perovskite materials.

Dr. Perini and Professor Correa-Baena discuss the latest research and effort to obtain higher performance and stability of perovskite materials.

Dr. Perini and Professor Correa-Baena discuss the latest research and effort to obtain higher performance and stability of perovskite materials.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service