Skip to Content
Merck
All Photos(4)

Key Documents

799386

Sigma-Aldrich

Potassium chlorate

anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KClO3
CAS Number:
Molecular Weight:
122.55
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
Assay:
≥99%
grade:
ACS reagent
anhydrous
form:
powder

grade

ACS reagent
anhydrous

Quality Level

product line

Redi-Dri

Assay

≥99%

form

powder

quality

free-flowing

impurities

≤0.001% N compounds
≤0.005% insolubles

pH

5-6.5 (25 °C, 61.3 g/L)

mp

356 °C (lit.)

anion traces

bromate (BrO3-): ≤0.015%
chloride (Cl-): ≤0.001%
sulfate (SO42-): passes test

cation traces

Ca: ≤0.002%
Fe: ≤3 ppm
Mg: ≤0.002%
Na: ≤0.01%
heavy metals: ≤5 ppm (by ICP)

SMILES string

[K+].[O-]Cl(=O)=O

InChI

1S/ClHO3.K/c2-1(3)4;/h(H,2,3,4);/q;+1/p-1

InChI key

VKJKEPKFPUWCAS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Legal Information

Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Flame over circleSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Ox. Sol. 1

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Carbon fragments are ripped off from graphite oxide sheets during their thermal reduction.
Jankovsky O, et al.
New. J. Chem., 38(12), 5700-5705 (2014)
A noncovalent compatibilization approach to improve the filler dispersion and properties of polyethylene/graphene composites.
Vasileiou AA, et al.
ACS Applied Materials & Interfaces, 6(3), 1916-1925 (2014)
ESR Spectrum of O-3 Trapped in a Single Crystal of Potassium Chlorate.
Schlick S.
J. Chem. Phys. , 56, 654-661 (1972)
Single sheet functionalized graphene by oxidation and thermal expansion of graphite.
McAllister MJ, et al.
Chemistry of Materials, 19(18), 4396-4404 (2007)
Daniel R Dreyer et al.
Chemical Society reviews, 39(1), 228-240 (2009-12-22)
The chemistry of graphene oxide is discussed in this critical review. Particular emphasis is directed toward the synthesis of graphene oxide, as well as its structure. Graphene oxide as a substrate for a variety of chemical transformations, including its reduction

Articles

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service