Skip to Content
Merck
All Photos(2)

Key Documents

796115

Sigma-Aldrich

Graphene dispersion

greener alternative

for gravure printing,with ethyl cellulose in terpineol, gravure printable

Synonym(s):

conductive ink

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352103
NACRES:
NA.23

product name

Graphene ink, for gravure printing, with ethyl cellulose in terpineol, gravure printable

Quality Level

form

liquid

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

resistivity

0.003-0.008 Ω-cm (thermally annealed 250 °C for 30 minutes, film thickness >100 nm)

particle size

≤3 μm

surface tension

30-35 dyn/cm

viscosity

0.75-3.00 Pa.s (shear viscosity at 100 s-1, 25 °C)

bp

213-218 °C (Terpineol)

density

0.9-1.1 g/mL at 25 °C

greener alternative category

SMILES string

OC(C1=C2C(C3=C(C4=C(C5=C(C6=C(C7=C(C8=C(C9=C(C%10=C(C%11=C(C%12=C(C%13=C(C%14=C(C%15=CC=C%16O)C%16=C%17)C%17=C%18)C%18=C%19C(O)=O)C%19=C%20)C%20=C%21O)C%21=C%22)C%22=C%23)C%23=C%24O)C%24=C%25)C%25=C%26C(O)=O)C%26=C%27)C%27=C2)=C%28C%29=C(C%30=C%31C%28=C%3

Looking for similar products? Visit Product Comparison Guide

General description

Gravure printable graphene dispersion has shown promising uses in printed and flexible electronics owning to its high speed and high resolution of deposition.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product belongs to Enabling category of greener alternatives thus aligns with "Design for energy efficency". 3D printable inks help to design and print more complex designs with minimum waste and it is environmentally friendly. Click here for more information.

Application

Formulated for Gravure printing.
Curing Condition: 250-350 °C, 20-30 min
This product can be used in material extrusion 3D printing technique.

Legal Information

Sold under Material Transfer Agreement with Mark Hersam group at Northwestern University. 

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rapid and Versatile Photonic Annealing of Graphene Inks for Flexible Printed Electronics
Ethan B. Secor , Bok Y. Ahn , Theodore Z. Gao , Jennifer A. Lewis , and Mark C. Hersam
Advanced Materials, 27, 6683-6688 (2015)
High?resolution patterning of graphene by screen printing with a silicon stencil for highly flexible printed electronics.
Hyun W J, et al.
Advanced Materials, 27(1), 109-115 (2015)
High-Performance Solid-State Supercapacitors and Microsupercapacitors Derived from Printable Graphene Inks
Lei Li, Ethan B. Secor, Kan-Sheng Chen, Jian Zhu, Xiaolong Liu, Theodore Z. Gao
Advanced Energy Materials, 6, N/A-N/A (2016)
Ethan B Secor et al.
Advanced materials (Deerfield Beach, Fla.), 26(26), 4533-4538 (2014-05-02)
Gravure printing of graphene is demonstrated for the rapid production of conductive patterns on flexible substrates. Development of suitable inks and printing parameters enables the fabrication of patterns with a resolution down to 30 μm. A mild annealing step yields

Articles

Functional materials for printed electronics applications enable flexible displays, RFID tags, and biomedical sensors.

Functional materials for printed electronics applications enable flexible displays, RFID tags, and biomedical sensors.

Functional materials for printed electronics applications enable flexible displays, RFID tags, and biomedical sensors.

Functional materials for printed electronics applications enable flexible displays, RFID tags, and biomedical sensors.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service