Skip to Content
Merck
All Photos(1)

Key Documents

776289

Sigma-Aldrich

N,N′-Bis(3-pentyl)perylene-3,4,9,10-bis(dicarboximide)

98% (HPLC)

Synonym(s):

2,9-Di(pent-3-yl)-anthra[2,1,9-def,6,5,10-d′e′f′]diisoquinoline-1,3,8,10-tetrone, N,N′-Bis(ethylpropyl)perylene-3,4,9,10-tetracarboxylicdimide, N,N-Bis(ethylpropyl)perylene-3,4,9,10-tetracarboxylicdiimide, Anthra[2,1,9-def:6,5,10-d′e′f′]diisoquinoline-1,3,8,10(2H,9H)-tetrone, 2,9-bis(1-ethylpropyl)-, PBI

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H30N2O4
CAS Number:
Molecular Weight:
530.61
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

98% (HPLC)

form

powder

mp

>450 °C

λmax

524, 488, 457 nm in dichloromethane

SMILES string

CCC(CC)N1C(=O)c2ccc3c4ccc5C(=O)N(C(CC)CC)C(=O)c6ccc(c7ccc(C1=O)c2c37)c4c56

InChI

1S/C34H30N2O4/c1-5-17(6-2)35-31(37)23-13-9-19-21-11-15-25-30-26(34(40)36(33(25)39)18(7-3)8-4)16-12-22(28(21)30)20-10-14-24(32(35)38)29(23)27(19)20/h9-18H,5-8H2,1-4H3

InChI key

AIPBSZJAQGGCPD-UHFFFAOYSA-N

General description

N,N′-Bis(3-pentyl)perylene-3,4,9,10-bis(dicarboximide) (EP-PDI) is a perylenediimide derivative that has a planar backbone and π-conjugation. It is a thermally stable polymer with luminescence and photochemical properties. It acts as a pigment and has charge mobility of 1 cm2V-1s-1 which can be used in a variety of energy and chemical sensing based applications.

Application

EP-PDI (acceptor) blended with P3HT (donor) and PCBM (donor) can be used in the fabrication of organic photovoltaic devices and organic solar cells. It forms a mixture with tetrahydrofuran (THF) that shows high intensity fluorescence which can be used as a sensor for the detection of nitroaromatic compounds.
High electron transporting character; Perylenebis(dicarboximide)s (PDIs) can be used as air-stable n-type semiconductor materials for organic fieldeffect transistors (OFETs);
Perylene bisimide dye used for organic photovoltaics (OPVs). A PCE of 1.75% was demonstrated with the solvent vapor annealed films; representing the best among the solar cells based on the single molecule materials.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Attilio Matera et al.
Foods (Basel, Switzerland), 9(4) (2020-04-05)
The quality of ready meals is affected by several factors that may impair stability and nutritional value. In this work, we evaluated the overall quality of four traditional meals (Basilicata region) prepared according to the cook&chill method, packaged in air
Neeraj K Surana et al.
Nature, 552(7684), 244-247 (2017-12-07)
Microbiome-wide association studies have established that numerous diseases are associated with changes in the microbiota. These studies typically generate a long list of commensals implicated as biomarkers of disease, with no clear relevance to disease pathogenesis. If the field is
Silvia Quattrosoldi et al.
Polymers, 12(1) (2020-01-23)
The crystallization behavior of fully biobased poly(butylene isophthalate) (PBI) has been investigated using calorimetric and microscopic techniques. PBI is an extremely slow crystallizing polymer that leads, after melt-crystallization, to the formation of lamellar crystals and rather large spherulites, due to
Ye Tian et al.
The Journal of experimental medicine, 214(11), 3347-3360 (2017-09-22)
T lymphocytes are key cellular components of the adaptive immune system and play a central role in cell-mediated immunity in vertebrates. Despite their heterogeneities, it is believed that all different types of T lymphocytes are generated exclusively via the differentiation
The effects of donor: acceptor intermolecular mixing and acceptor crystallization on the composition ratio of blended, spin coated organic thin films.
Weintraub MT, et al.
Journal of Material Chemistry C, 4(33), 7756-7765 (2016)

Articles

Fabrication procedure of organic field effect transistor device using a soluble pentacene precursor.

Solution-processed organic photovoltaic devices (OPVs) have emerged as a promising clean energy generating technology due to their ease of fabrication, potential to enable low-cost manufacturing via printing or coating techniques, and ability to be incorporated onto light weight, flexible substrates.

Solution-processed organic photovoltaic devices (OPVs) have emerged as a promising clean energy generating technology due to their ease of fabrication, potential to enable low-cost manufacturing via printing or coating techniques, and ability to be incorporated onto light weight, flexible substrates.

Solution-processed organic photovoltaic devices (OPVs) have emerged as a promising clean energy generating technology due to their ease of fabrication, potential to enable low-cost manufacturing via printing or coating techniques, and ability to be incorporated onto light weight, flexible substrates.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service