Skip to Content
Merck
All Photos(3)

Key Documents

75992

Sigma-Aldrich

Palladium on activated charcoal

greener alternative

5% Pd basis

Synonym(s):

Pd/C

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Pd
MDL number:
UNSPSC Code:
12141733
eCl@ss:
38191001
PubChem Substance ID:
NACRES:
NA.22

form

powder

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst

greener alternative product characteristics

Designing Safer Chemicals
Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

concentration

5% Pd

greener alternative category

SMILES string

[Pd]

InChI

1S/Pd

InChI key

KDLHZDBZIXYQEI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Palladium on activated charcoal (Pd/C) is a catalyst used in hydrogenation of alkenes, alkynes, ketones, nitriles, imines, nitro groups, aromatic and heteroaromatic systems. Pd/C also catalyzes phosphination of aryl bromides and triflates with triphenylphines to yield aryl phosphines.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for catalytic efficiency. Click here for more information.

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sonogashira coupling of aryl halides catalyzed by palladium on charcoal.
Novak, Zoltan et al.
The Journal of Organic Chemistry, 68(8), 3327-3329 (2003)
Synthesis of aryl phosphines by phosphination with triphenylphosphine catalyzed by palladium on charcoal.
Lai, Chi Wai et al.
Tetrahedron Letters, 42(29), 4883-4885 (2001)
Examination of the aromatic amination catalyzed by palladium on charcoal.
Komaromi, Anna and Novak, Zoltan
advanced synthesis and catalysis, 352(9), 1523-1532 (2010)
Palladium on Carbon.
eEROS (Encyclopedia of Reagents for Organic Synthesis) (2001)
Examination of the aromatic amination catalyzed by palladium on charcoal.
Komaromi, Anna and Novak, Zoltan
Advanced Synthesis & Catalysis, 352(9), 1523-1532 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service